Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01306674_m1
          See other FAM159B GE Assays ›
          Gene Symbol:
          Fam159b
          Gene Name:
          family with sequence similarity 159, member B
          Gene Aliases:
          A930021C24Rik
          Chromosome Location:
          Chr.13: 104845283 - 104863893 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01306674_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam159b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029984.1 NP_084260.1 2 - 3 532 69
          GenBank mRNA AK020882.1 - 2 - 3 396 69
          AK042897.1 - 2 - 3 532 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002605.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fam159b

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29292939 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29292940 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135788 NM_029984.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135788 AK042897.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 686944 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 739630 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 425675 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 334940 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 371198 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 238160 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 665377 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 475269 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 494292 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 90425 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 408332 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 410142 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 380600 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 666383 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 619934 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 121381 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 62426 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 125960 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 17065 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33802 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 86996 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139865 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166537 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135788 NM_029984.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135788 AK042897.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29292952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29292932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29292946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29292963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29292947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29292930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29292937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29292957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29292950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29292935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29292939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29292954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29292942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29292926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29292928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29292933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29292944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29292961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29292956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29292965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29292922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29292925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29292941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29292924 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02766351 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07982532 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139865 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166537 - -
          Whole Mouse Genome G4122A A_52_P188295 NM_029984.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P188295 AK020882.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P188295 AK042897.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139865 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07982532 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166537 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10609678 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10609679 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10609674 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29292940 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29292939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29292809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10786268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10596790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10542845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10969402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10526091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10937986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10623950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10739062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10514115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10786271 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10764789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29292907 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10411948 - -
          Affymetrix Mouse Gene 1.0 ST Array 10411946 - -
          Affymetrix Mouse Gene 1.0 ST Array 10411947 - -
          Affymetrix HT MG-430 PM Array Plate 1430977_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10411945 - -
          Mouse Genome 430 2.0 Array 1430977_at - -
          Mouse Genome 430 2.0 Array 1430977_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77803_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77803_at - -
          Affymetrix Mouse Clariom S Array PSR1300017745.mm.1 NM_029984.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017745.mm.1 NM_029984.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017745.mm.1 TC1300002605.mm.1 MTA TC
          Mouse Expression Set 430 1430977_at - -
          Mouse Expression Set 430 1430977_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430977_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430977_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 726407 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1213789 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1213789 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3124611 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3048751 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3048751 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3124611 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline