Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01310297_g1
          See other DIMT1 GE Assays ›
          Gene Symbol:
          Dimt1
          Gene Name:
          DIM1 dimethyladenosine transferase 1-like (S. cerevisiae)
          Gene Aliases:
          1500031M22Rik, AV111347, Dimt1l
          Chromosome Location:
          Chr.13: 106947043 - 106960225 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01310297_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dimt1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025447.4 NP_079723.1 1 - 2 154 54
          GenBank mRNA AK007759.1 - 1 - 2 105 54
          AK011544.1 - 1 - 2 111 54
          AK015145.1 - 1 - 2 104 54
          AK078186.1 - 1 - 2 106 54
          AK078401.1 - 1 - 2 128 54
          AK146188.1 - 1 - 2 125 54
          AK161333.1 - 1 - 2 106 54
          BC019799.1 - 1 - 2 127 5053500 54
          BF608861.1 - 1 - 2 165 54

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
          This assay is in the 5'UTR region of the AK078186.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dimt1

          Panther Classification:

          Molecular Function -

          RNA methyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          rRNA modification
          rRNA processing
          methylation
          rRNA (adenine-N6,N6-)-dimethyltransferase activity
          RNA binding
          methyltransferase activity
          rRNA methyltransferase activity
          transferase activity
          poly(A) RNA binding
          18S rRNA (adenine(1779)-N(6)/adenine(1780)-N(6))-dimethyltransferase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1300007546.mm.1 NM_025447.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007546.mm.1 NM_025447.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007546.mm.1 TC1300001190.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300007544.mm.1 NM_025447.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007544.mm.1 NM_025447.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007544.mm.1 TC1300001190.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300007550.mm.1 NM_025447.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007550.mm.1 NM_025447.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007550.mm.1 TC1300001190.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300007548.mm.1 NM_025447.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007548.mm.1 NM_025447.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007548.mm.1 TC1300001190.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300007549.mm.1 NM_025447.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007549.mm.1 NM_025447.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007549.mm.1 TC1300001190.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300007545.mm.1 NM_025447.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007545.mm.1 NM_025447.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007545.mm.1 TC1300001190.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300007547.mm.1 NM_025447.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007547.mm.1 NM_025447.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007547.mm.1 TC1300001190.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300007551.mm.1 NM_025447.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007551.mm.1 NM_025447.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007551.mm.1 TC1300001190.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 475408 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline