Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01310378_m1
          See other NKIRAS1 GE Assays ›
          Gene Symbol:
          Nkiras1
          Gene Name:
          NFKB inhibitor interacting Ras-like protein 1
          Gene Aliases:
          2400004O09Rik, KBRAS1, kappaB-Ras1
          Chromosome Location:
          Chr.14: 18271142 - 18284003 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01310378_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nkiras1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023526.3 NP_076015.2 3 - 4 540 95
          GenBank mRNA AF225707.1 - 3 - 4 397 95
          AK010285.1 - 4 - 5 713 95
          AK028539.1 - 3 - 4 528 95
          AK148040.1 - 3 - 4 466 95
          AK167340.1 - 3 - 4 437 95
          BC061092.1 - 3 - 4 445 6589347 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400000141.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nkiras1

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          small GTPase mediated signal transduction
          biological_process
          nucleotide binding
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29437793 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29437789 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29437791 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P448391 BC061092.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P448391 AF225707.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P448391 AK010285.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P448391 AK028539.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P448391 AK148040.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P448391 BC061092.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P448391 AF225707.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P448391 AK010285.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P448391 AK028539.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P448391 AK148040.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 565069 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 387474 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 608012 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 858371 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 668342 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 269414 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233523 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 276393 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 348766 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 685243 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 132086 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P144246 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146439 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P448391 BC061092.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P448391 AF225707.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P448391 AK010285.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P448391 AK028539.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P448391 AK148040.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P448391 BC061092.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P448391 AF225707.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P448391 AK010285.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P448391 AK028539.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P448391 AK148040.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P448391 BC061092.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P448391 AF225707.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P448391 AK010285.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P448391 AK028539.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P448391 AK148040.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P448391 BC061092.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P448391 AF225707.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P448391 AK010285.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P448391 AK028539.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P448391 AK148040.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29437801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29437794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29437804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29437795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29437799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29437797 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29437806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29437800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29437808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29437789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29437790 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146439 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08041929 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P144246 - -
          Whole Mouse Genome G4122A A_52_P52982 AK028539.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P52982 NM_023526.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P448391 BC061092.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P448391 AF225707.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P448391 AK010285.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P448391 AK028539.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P448391 AK148040.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P448391 BC061092.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P448391 AF225707.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P448391 AK010285.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P448391 AK028539.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P448391 AK148040.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117697 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146439 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P144246 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11167378 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11167383 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11167345 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29437791 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29437789 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29437793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29437855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29437821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29437859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29437820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29437797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29437795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29437801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29437789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11167388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29437825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29437818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29437815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29437807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11629953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29437813 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29437800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11767482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29437824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29437804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29437799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29437812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29437802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11337821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29437823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29437857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29437794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29437811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29437808 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10412903 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412902 - -
          Affymetrix Mouse Gene 1.0 ST Array 10417750 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412906 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412901 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412904 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412905 - -
          Affymetrix HT MG-430 PM Array Plate 1428503_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10417749 - -
          Affymetrix Mouse Gene 1.1 ST Array 10412900 - -
          Mouse Genome 430 2.0 Array 1428503_a_at - -
          Mouse Genome 430 2.0 Array 1428503_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69721_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69721_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1400001154.mm.1 NM_023526.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001154.mm.1 TC1400000141.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400001157.mm.1 NM_023526.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001157.mm.1 TC1400000141.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400001158.mm.1 NM_023526.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001158.mm.1 TC1400000141.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400001155.mm.1 NM_023526.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400001155.mm.1 TC1400000141.mm.1 MTA TC
          Mouse Expression Set 430 1428503_PM_a_at - -
          Mouse Expression Set 430 1428503_a_at - -
          Murine Genome U74v2 Set 160991_at - -
          Murine Genome U74v2 Set 160991_PM_at - -
          Mouse Genome 430A 2.0 Array 1428503_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1428503_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1428503_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1428503_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 836589 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5570671 - -
          Illumina mouse-6 v1.0 expression beadchip 2510440 - -
          Illumina mouse-6 v1.0 expression beadchip 5570671 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002510440 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005570671 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2656189 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1238326 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2649696 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1238326 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2649696 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2656189 - -
          mouse-6 v1.1 (Illumina) scl069721.4_6-S - -
          mouse-6 v1.1 (Illumina) scl0069721.2_295-S - -
          Illumina Sentrix mouse-6.v1 bead array 2510440 - -
          Illumina Sentrix mouse-6.v1 bead array 5570671 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl069721.4_6-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0069721.2_295-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5570671 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2510440 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2649696 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2656189 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2649696 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2656189 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline