Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01317328_m1
          See other NEGR1 GE Assays ›
          Gene Symbol:
          Negr1
          Gene Name:
          neuronal growth regulator 1
          Gene Aliases:
          5330422G01Rik, KILON, Ntra
          Chromosome Location:
          Chr.3: 156561794 - 157316464 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01317328_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Negr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039094.3 NP_001034183.1 1 - 2 559 66
          NM_177274.4 NP_796248.1 1 - 2 559 66
          GenBank mRNA AJ487032.1 - 1 - 2 179 66
          AK030503.1 - 1 - 2 385 66
          AK035218.1 - 1 - 2 395 66
          AK142298.1 - 1 - 2 253 66
          AK147253.1 - 1 - 2 560 66
          BC146005.1 - 1 - 2 344 40131308 66
          BC146007.1 - 1 - 2 343 40131310 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300001562.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Negr1

          Panther Classification:

          Molecular Function -

          immunoglobulin

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          locomotory behavior
          feeding behavior
          positive regulation of neuron projection development
          single organismal cell-cell adhesion
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300012611.mm.1 NM_001039094.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300012611.mm.1 NM_177274.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300012611.mm.1 TC0300001562.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300012613.mm.1 NM_001039094.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300012613.mm.1 NM_177274.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300012613.mm.1 TC0300001562.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300012608.mm.1 NM_001039094.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300012608.mm.1 NM_177274.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300012608.mm.1 TC0300001562.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300012616.mm.1 NM_001039094.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300012616.mm.1 NM_177274.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300012616.mm.1 TC0300001562.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300012610.mm.1 NM_001039094.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300012610.mm.1 NM_177274.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300012610.mm.1 TC0300001562.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300012617.mm.1 NM_001039094.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300012617.mm.1 NM_177274.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300012617.mm.1 TC0300001562.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline