Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01321394_m1
          See other ADH6A GE Assays ›
          Gene Symbol:
          Adh6a
          Gene Name:
          alcohol dehydrogenase 6A (class V)
          Gene Aliases:
          1810009C16Rik, Adh5a
          Chromosome Location:
          Chr.3: 138313286 - 138331134 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01321394_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adh6a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026945.1 NP_081221.1 7 - 8 1021 132
          GenBank mRNA AK007397.1 - 7 - 8 1017 132
          AK028114.1 - 7 - 8 1023 133
          BC130016.1 - 7 - 8 990 40140468 132

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300001410.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          dehydrogenase oxidoreductase metabolite interconversion enzyme

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2112882 AK007397.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2112882 NM_026945.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 296605 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233190 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 828924 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 625196 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 361778 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 932694 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 876209 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 174316 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 141127 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 134626 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 166300 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165191 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140090 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2112882 AK007397.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2112882 NM_026945.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165191 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140090 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05090334 - -
          Whole Mouse Genome G4122A A_52_P276348 AK007397.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P276348 AK028114.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P276348 BC130016.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P276348 NM_026945.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P299608 AK028114.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140090 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22568260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568156 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568224 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568147 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02742858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568195 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03040506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02989712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568157 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568163 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568199 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03129943 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568155 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02432472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568148 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568189 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03001825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03001823 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568158 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568149 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568166 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03001811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02432478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03154050 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568143 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03001826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568197 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568152 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02432458 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22568178 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10496452 - -
          Affymetrix Mouse Gene 1.0 ST Array 10496454 - -
          Affymetrix Mouse Gene 1.0 ST Array 10496448 - -
          Affymetrix Mouse Gene 1.0 ST Array 10496453 - -
          Affymetrix Mouse Gene 1.0 ST Array 10496449 - -
          Affymetrix Mouse Gene 1.0 ST Array 10496450 - -
          Affymetrix Mouse Gene 1.0 ST Array 10496451 - -
          Affymetrix HT MG-430 PM Array Plate 1429608_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10496447 - -
          Mouse Genome 430 2.0 Array 1429608_at - -
          Mouse Genome 430 2.0 Array 1429608_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69117_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69117_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0300011390.mm.1 NM_026945.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300011390.mm.1 NM_026945.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300011390.mm.1 TC0300001410.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300011389.mm.1 NM_026945.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300011389.mm.1 NM_026945.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300011389.mm.1 TC0300001410.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300011393.mm.1 NM_026945.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300011393.mm.1 NM_026945.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300011393.mm.1 TC0300001410.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300011392.mm.1 NM_026945.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300011392.mm.1 NM_026945.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300011392.mm.1 TC0300001410.mm.1 MTA TC
          Mouse Expression Set 430 1429608_PM_at - -
          Mouse Expression Set 430 1429608_at - -
          Affymetrix HT Mouse Genome 430B Array 1429608_at - -
          Affymetrix HT Mouse Genome 430B Array 1429608_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 369990 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4060463 - -
          Illumina mouse-6 v1.0 expression beadchip 104670577 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1247628 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247628 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223481 - -
          Illumina Sentrix mouse-6.v1 bead array 104670577 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104670577 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1223481 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline