Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01324837_m1
          See other SMIM13 GE Assays ›
          Gene Symbol:
          Smim13
          Gene Name:
          small integral membrane protein 13
          Gene Aliases:
          2900036K02Rik
          Chromosome Location:
          Chr.13: 41249844 - 41276577 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01324837_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smim13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001135577.2 NP_001129049.1 1 - 2 468 91
          GenBank mRNA AK082625.1 - 1 - 2 75 91
          BC024659.1 - 1 - 2 140 91
          BC066054.1 - 1 - 2 452 91
          BC094222.1 - 1 - 2 452 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000466.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P123938 NM_001135577.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P123938 AK076140.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P123938 BC066054.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P123938 BC094222.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P123938 BE949208.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 790287 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 389543 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 113408 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 750732 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 97328 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 29331 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 918204 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 721139 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 450497 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 416450 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 761125 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 43973 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 489626 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 459956 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 635393 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 161091 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 173939 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166895 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P123938 NM_001135577.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P123938 AK076140.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P123938 BC066054.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P123938 BC094222.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P123938 BE949208.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166895 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07849601 - -
          Whole Mouse Genome G4122A A_52_P239742 NM_001135577.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P239742 AK082625.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P239742 BC024659.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P239742 BC066054.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P239742 BC094222.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P674568 NM_001135577.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P674568 BC024659.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P674568 BC066054.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P674568 BC094222.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123938 NM_001135577.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123938 AK076140.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123938 BC066054.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123938 BC094222.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123938 BE949208.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11095355 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28976916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28976932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28976929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28976940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28976944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28976918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28977132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28977121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28976913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28977124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28976947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28977130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28977125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28976942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28977117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28976930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28977128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28977115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28976911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11087630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10888160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28976927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28976938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28976922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10810902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28976912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28976924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28976914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10972304 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28976933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28977126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10751993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28977119 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1433986_PM_at - -
          Mouse Genome 430 2.0 Array 1433986_at - -
          Mouse Genome 430 2.0 Array 1433986_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 108934_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 108934_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300002814.mm.1 NM_001135577.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002814.mm.1 NM_001135577.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002814.mm.1 TC1300000466.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300002813.mm.1 NM_001135577.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002813.mm.1 NM_001135577.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002813.mm.1 TC1300000466.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300002815.mm.1 NM_001135577.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002815.mm.1 NM_001135577.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002815.mm.1 TC1300000466.mm.1 MTA TC
          Affymetrix HT Mouse Genome 430B Array 1433986_at - -
          Affymetrix HT Mouse Genome 430B Array 1433986_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 310766 - -
          ABI Mouse Genome Survey Microarray 530309 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline