Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01327811_m1
          See other CERCAM GE Assays ›
          Gene Symbol:
          Cercam
          Gene Name:
          cerebral endothelial cell adhesion molecule
          Gene Aliases:
          2600006K01Rik, AL024097, Ceecam1
          Chromosome Location:
          Chr.2: 29845105 - 29882845 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01327811_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cercam
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207298.2 NP_997181.1 8 - 9 1110 64
          GenBank mRNA AK155384.1 - 8 - 9 1106 64
          BC052176.1 - 8 - 9 1073 64
          BC138847.1 - 8 - 9 1089 8861869 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000514.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cercam

          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21099938 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21099935 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21099934 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21099936 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P559545 BC138847.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P559545 AK155384.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P559545 BC052176.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P559545 NM_207298.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P559545 BC138847.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P559545 AK155384.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P559545 BC052176.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P559545 NM_207298.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 40305 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 177041 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 397786 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 118976 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 333334 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 388046 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 152810 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126333 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123678 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P559545 BC138847.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P559545 AK155384.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P559545 BC052176.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P559545 NM_207298.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P559545 BC138847.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P559545 AK155384.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P559545 BC052176.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P559545 NM_207298.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21099937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21099953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21099938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21099925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21099936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21099940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21099933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21099942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21099950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21099928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21099955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21099920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21099930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21099956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21099952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21099921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21099946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21099945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21099929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21099923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21099943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21099948 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123678 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00333239 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126333 - -
          Whole Mouse Genome G4122A A_52_P559545 BC138847.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P559545 AK155384.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P559545 BC052176.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P559545 NM_207298.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P559545 BC138847.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P559545 AK155384.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P559545 BC052176.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P559545 NM_207298.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121136 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104398 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123678 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126333 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01454692 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01152009 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01152008 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01152013 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01151997 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21099935 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21099934 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21099936 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21099938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21100060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21099946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099917 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21100013 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01889653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099939 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01417855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01763092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02177497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099914 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099943 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01763095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01835492 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02177498 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01186009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01152009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02276867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01194088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099956 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01287391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01806375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099948 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099969 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01766760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099942 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01417857 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21099901 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10481382 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470782 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470780 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470781 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470783 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470777 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470787 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470778 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470785 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470776 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470784 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470786 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470779 - -
          Affymetrix HT MG-430 PM Array Plate 1435345_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10470775 - -
          Affymetrix Mouse Gene 1.1 ST Array 10481380 - -
          Mouse Genome 430 2.0 Array 1432448_PM_at - -
          Mouse Genome 430 2.0 Array 1435345_at - -
          Mouse Genome 430 2.0 Array 1435345_PM_at - -
          Mouse Genome 430 2.0 Array 1432448_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 99151_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 99151_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200003808.mm.1 NM_207298.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200003808.mm.1 NM_207298.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200003808.mm.1 TC0200000514.mm.1 MTA TC
          Mouse Expression Set 430 1435345_PM_at - -
          Mouse Expression Set 430 1435345_at - -
          Affymetrix HT Mouse Genome 430B Array 1435345_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435345_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 823615 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 780605 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000780605 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2671755 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2671755 - -
          mouse-6 v1.1 (Illumina) scl0099151.1_201-S - -
          Illumina Sentrix mouse-6.v1 bead array 780605 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0099151.1_201-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 780605 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2671755 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2671755 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0099151.1_201-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline