Search
Search
| Gene Symbol: Ell3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145973.2 | NP_666085.2 | 6 - 7 | 694 | 66 | |
| GenBank mRNA | AK139676.1 | - | 7 - 8 | 1120 | 66 | |
| BC045151.1 | - | 6 - 7 | 694 | 6313358 | 66 | |
Set Membership: |
|