Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01331643_m1
          See other TEX22 GE Assays ›
          Gene Symbol:
          Tex22
          Gene Name:
          testis expressed gene 22
          Gene Aliases:
          1700028O09Rik, AV210422, Tep22
          Chromosome Location:
          Chr.12: 113074469 - 113088917 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01331643_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tex22
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029381.1 NP_083657.1 2 - 3 834 60
          GenBank mRNA AF522069.1 - 2 - 3 834 60
          AK006471.1 - 2 - 3 302 60
          BC048634.1 - 2 - 3 267 6770610 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200001279.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tex22

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087399 AK006471.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2134064 AF522069.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2134064 NM_029381.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 535518 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 536101 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 322656 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 466788 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 920734 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 547987 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 158426 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 294347 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 362520 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 5770 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 164140 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133596 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087399 AK006471.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2134064 AF522069.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2134064 NM_029381.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P282286 AK006471.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P282286 BC048634.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09985031 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09985029 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09985042 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09985030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28754587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28754588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28754583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28754580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28754582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28754590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28754592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28754586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28754577 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10398968 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398967 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398971 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398970 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398969 - -
          Affymetrix HT MG-430 PM Array Plate 1429548_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10398966 - -
          Mouse Genome 430 2.0 Array 1429548_at - -
          Mouse Genome 430 2.0 Array 1429548_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75671_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75671_at - -
          Affymetrix Mouse Clariom S Array PSR1200009233.mm.1 NM_029381.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009233.mm.1 NM_029381.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009233.mm.1 TC1200001279.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200009228.mm.1 NM_029381.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009228.mm.1 NM_029381.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009228.mm.1 TC1200001279.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200009224.mm.1 NM_029381.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009224.mm.1 NM_029381.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009224.mm.1 TC1200001279.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200009230.mm.1 NM_029381.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009230.mm.1 NM_029381.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009230.mm.1 TC1200001279.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200009232.mm.1 NM_029381.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009232.mm.1 NM_029381.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009232.mm.1 TC1200001279.mm.1 MTA TC
          Mouse Expression Set 430 1429548_at - -
          Mouse Expression Set 430 1429548_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429548_at - -
          Affymetrix HT Mouse Genome 430B Array 1429548_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 380045 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2652235 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2652235 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline