Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01336392_m1
          See other ENTPD8 GE Assays ›
          Gene Symbol:
          Entpd8
          Gene Name:
          ectonucleoside triphosphate diphosphohydrolase 8
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 25080146 - 25085719 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01336392_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Entpd8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028093.1 NP_082369.1 3 - 4 492 64
          GenBank mRNA AY364442.1 - 3 - 4 354 64
          BC031143.1 - 3 - 4 488 4973216 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000397.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          hydrolase nucleotide phosphatase phosphatase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleoside monophosphate biosynthetic process
          nucleoside diphosphate biosynthetic process
          nucleotide binding
          ATP binding
          hydrolase activity
          nucleoside-diphosphatase activity
          nucleoside-triphosphatase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989215 NM_028093.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989215 AV026718.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 636113 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 480354 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 926089 AF309644.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 926089 AK003118.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 926089 BC057975.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 926089 NM_008993.2 refseq_mrna_mouse_NM
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P180571 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989215 NM_028093.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989215 AV026718.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468482 BC031143.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468482 NM_028093.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468482 AK008590.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468482 AV026718.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P468482 AY364442.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P180571 - -
          Whole Mouse Genome G4122A A_51_P468482 BC031143.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468482 NM_028093.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468482 AK008590.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468482 AV026718.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468482 AY364442.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P180571 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01314902 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01314892 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01314889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21069713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21069690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01979239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01166868 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21069717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21069681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21069678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21069672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21069715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21069712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01887210 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21069704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21069725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01887213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01314902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01166869 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21069720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01979238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01225177 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21069714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01166861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21069707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21069713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21069677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01887218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21069679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21069684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21069701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01887211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21069709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21069699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01166838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21069721 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21069683 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21069688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01879188 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10469935 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469931 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469926 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469930 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469927 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469932 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469924 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469925 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469933 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469928 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469929 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469934 - -
          Affymetrix HT MG-430 PM Array Plate 1429550_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10480585 - -
          Affymetrix Mouse Gene 1.1 ST Array 10469923 - -
          Mouse Genome 430 2.0 Array 1429550_PM_at - -
          Mouse Genome 430 2.0 Array 1429550_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72090_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72090_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200002178.mm.1 NM_028093.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002178.mm.1 NM_028093.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002178.mm.1 TC0200000397.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200002179.mm.1 NM_028093.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002179.mm.1 NM_028093.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002179.mm.1 TC0200000397.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200002180.mm.1 NM_028093.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002180.mm.1 NM_028093.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002180.mm.1 TC0200000397.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200002181.mm.1 NM_028093.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002181.mm.1 NM_028093.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002181.mm.1 TC0200000397.mm.1 MTA TC
          Mouse Expression Set 430 1429550_PM_at - -
          Mouse Expression Set 430 1429550_at - -
          Murine Genome U74v2 Set 165683_at - -
          Murine Genome U74v2 Set 165683_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429550_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429550_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 407466 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4920079 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2744998 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2744998 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2744998 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline