Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01337414_gH
          See other GTF2A1 GE Assays ›
          Gene Symbol:
          Gtf2a1
          Gene Name:
          general transcription factor II A, 1
          Gene Aliases:
          6330549H03Rik, AA536742, AA959775, AW060250, TfIIAa/b, Tfiia1
          Chromosome Location:
          Chr.12: 91555262 - 91590487 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01337414_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gtf2a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031391.2 NP_113568.2 6 - 7 706 65
          NM_175335.3 NP_780544.1 6 - 7 693 65
          GenBank mRNA AF250834.1 - 6 - 7 636 65
          AK048711.1 - 6 - 7 694 65
          AK139996.1 - 6 - 7 706 65
          BC079909.1 - 6 - 7 639 30658355 65
          BC117829.1 - 6 - 7 663 40092917 65
          BC117830.1 - 6 - 7 663 40092921 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200002152.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gtf2a1

          Panther Classification:

          Molecular Function -

          general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          RNA polymerase II repressing transcription factor binding
          DNA binding
          transcription coactivator activity
          protein binding
          transcription factor binding
          TBP-class protein binding
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200014954.mm.1 NM_031391.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014954.mm.1 NM_031391.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014954.mm.1 NM_175335.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014954.mm.1 NM_175335.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014954.mm.1 TC1200002152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014966.mm.1 NM_031391.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014966.mm.1 NM_031391.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014966.mm.1 TC1200002152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014953.mm.1 NM_031391.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014953.mm.1 NM_031391.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014953.mm.1 NM_175335.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014953.mm.1 NM_175335.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014953.mm.1 TC1200002152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014960.mm.1 NM_031391.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014960.mm.1 NM_031391.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014960.mm.1 NM_175335.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014960.mm.1 NM_175335.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014960.mm.1 TC1200002152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014961.mm.1 NM_031391.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014961.mm.1 NM_031391.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014961.mm.1 NM_175335.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014961.mm.1 NM_175335.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014961.mm.1 TC1200002152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014962.mm.1 NM_031391.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014962.mm.1 NM_031391.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014962.mm.1 NM_175335.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014962.mm.1 NM_175335.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014962.mm.1 TC1200002152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014950.mm.1 NM_031391.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014950.mm.1 NM_031391.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014950.mm.1 NM_175335.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014950.mm.1 NM_175335.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014950.mm.1 TC1200002152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014957.mm.1 NM_031391.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014957.mm.1 NM_031391.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014957.mm.1 NM_175335.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014957.mm.1 NM_175335.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014957.mm.1 TC1200002152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014963.mm.1 NM_031391.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014963.mm.1 NM_031391.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014963.mm.1 NM_175335.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014963.mm.1 NM_175335.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014963.mm.1 TC1200002152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014970.mm.1 NM_175335.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014970.mm.1 NM_175335.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014970.mm.1 TC1200002152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014952.mm.1 NM_031391.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014952.mm.1 NM_031391.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014952.mm.1 NM_175335.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014952.mm.1 NM_175335.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014952.mm.1 TC1200002152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014955.mm.1 NM_031391.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014955.mm.1 NM_031391.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014955.mm.1 NM_175335.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014955.mm.1 NM_175335.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014955.mm.1 TC1200002152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014971.mm.1 NM_175335.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014971.mm.1 NM_175335.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014971.mm.1 TC1200002152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014956.mm.1 NM_031391.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014956.mm.1 NM_031391.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014956.mm.1 NM_175335.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014956.mm.1 NM_175335.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014956.mm.1 TC1200002152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014958.mm.1 NM_031391.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014958.mm.1 NM_031391.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014958.mm.1 NM_175335.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014958.mm.1 NM_175335.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014958.mm.1 TC1200002152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014959.mm.1 NM_031391.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014959.mm.1 NM_031391.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014959.mm.1 NM_175335.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014959.mm.1 NM_175335.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014959.mm.1 TC1200002152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014965.mm.1 NM_031391.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014965.mm.1 NM_031391.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014965.mm.1 NM_175335.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014965.mm.1 NM_175335.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014965.mm.1 TC1200002152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200014967.mm.1 NM_031391.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014967.mm.1 NM_031391.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200014967.mm.1 TC1200002152.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline