Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01339551_m1
          See other TMCO5B GE Assays ›
          Gene Symbol:
          Tmco5b
          Gene Name:
          transmembrane and coiled-coil domains 5B
          Gene Aliases:
          4930563P21Rik
          Chromosome Location:
          Chr.2: 113285095 - 113297190 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01339551_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmco5b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029232.2 NP_083508.1 11 - 12 856 146
          GenBank mRNA AK016214.1 - 11 - 12 847 146
          BC050803.1 - 11 - 12 856 6771315 146

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001680.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmco5b

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P130556 AK016214.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P130556 AK016214.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 702822 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 54595 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 272819 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 918676 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 496821 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 355555 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 484865 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 111706 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 127672 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167995 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P144935 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P130556 AK016214.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P130556 AK016214.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21533587 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167995 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P144935 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04650461 - -
          Whole Mouse Genome G4122A A_51_P130556 AK016214.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P130556 AK016214.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P144935 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167995 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01211815 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01211814 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01211817 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01211821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21533643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01181122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02246788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02267744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533604 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01255205 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01636710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533552 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01636711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02275489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02249953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533600 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01211811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533591 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02275492 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02246799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01211821 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02196029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02179410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533564 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533571 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01636713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01636700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01636712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02275487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02275493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02275503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01211815 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533588 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533601 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01211814 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02275496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02275502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02074012 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21533608 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10474609 - -
          Affymetrix Mouse Gene 1.0 ST Array 10474618 - -
          Affymetrix Mouse Gene 1.0 ST Array 10474607 - -
          Affymetrix Mouse Gene 1.0 ST Array 10474614 - -
          Affymetrix Mouse Gene 1.0 ST Array 10474615 - -
          Affymetrix Mouse Gene 1.0 ST Array 10474616 - -
          Affymetrix Mouse Gene 1.0 ST Array 10474610 - -
          Affymetrix Mouse Gene 1.0 ST Array 10474613 - -
          Affymetrix Mouse Gene 1.0 ST Array 10474612 - -
          Affymetrix Mouse Gene 1.0 ST Array 10474617 - -
          Affymetrix Mouse Gene 1.0 ST Array 10474611 - -
          Affymetrix Mouse Gene 1.0 ST Array 10474608 - -
          Affymetrix HT MG-430 PM Array Plate 1432203_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10474606 - -
          Mouse Genome 430 2.0 Array 1432203_PM_at - -
          Mouse Genome 430 2.0 Array 1432203_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75275_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75275_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200012480.mm.1 NM_029232.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200012480.mm.1 NM_029232.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200012480.mm.1 TC0200001680.mm.1 MTA TC
          Mouse Expression Set 430 1432203_at - -
          Mouse Expression Set 430 1432203_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432203_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432203_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 532444 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1255963 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1255963 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline