Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01340444_m1
          See other DDA1 GE Assays ›
          Gene Symbol:
          Dda1
          Gene Name:
          DET1 and DDB1 associated 1
          Gene Aliases:
          1500034J01Rik, 1700095J19Rik, R74921
          Chromosome Location:
          Chr.8: 71469171 - 71476097 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01340444_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dda1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001294259.1 NP_001281188.1 3 - 4 252 58
          NM_025600.2 NP_079876.1 4 - 5 333 58
          NR_122114.1 3 - 4 281 58
          NR_122115.1 4 - 5 329 58
          GenBank mRNA AK006318.1 - 4 - 5 317 58
          AK007085.1 - 4 - 5 352 58
          AK032966.1 - 4 - 5 291 58
          AK149117.1 - 3 - 4 263 58
          AK149980.1 - 4 - 5 313 58
          AK160132.1 - 4 - 5 330 58
          AK168085.1 - 4 - 5 291 58
          BB610194.1 - 4 - 5 326 58
          BC034701.1 - 4 - 5 293 3494430 58
          BC093523.1 - 4 - 5 276 3711509 58
          BY297077.1 - 3 - 4 212 58
          BY316490.1 - 4 - 5 333 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000791.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P407854 BC034701.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P407854 BC079637.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P407854 BC093523.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P407854 AK149980.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P407854 AK160132.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P407854 BC034701.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P407854 BC079637.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P407854 BC093523.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P407854 AK149980.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P407854 AK160132.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164495 BC034701.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164495 BC069957.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164495 BC079637.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164495 BC093523.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164495 AK149117.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164495 BC034701.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164495 BC069957.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164495 BC079637.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164495 BC093523.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164495 AK149117.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 197066 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 265570 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 910428 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 689400 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 10451 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179311 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P407854 BC034701.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P407854 BC079637.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P407854 BC093523.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P407854 AK149980.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P407854 AK160132.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P407854 BC034701.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P407854 BC079637.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P407854 BC093523.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P407854 AK149980.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P407854 AK160132.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164495 BC034701.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164495 BC069957.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164495 BC079637.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164495 BC093523.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164495 AK149117.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164495 BC034701.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164495 BC069957.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164495 BC079637.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164495 BC093523.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164495 AK149117.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179311 - -
          Whole Mouse Genome G4122A A_52_P407854 BC034701.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P407854 BC079637.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P407854 BC093523.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P407854 AK149980.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P407854 AK160132.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P407854 BC034701.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P407854 BC079637.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P407854 BC093523.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P407854 AK149980.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P407854 AK160132.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1125218 - -
          Whole Mouse Genome G4122A A_51_P164495 BC034701.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164495 BC069957.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164495 BC079637.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164495 BC093523.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164495 AK149117.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164495 BC034701.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164495 BC069957.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164495 BC079637.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164495 BC093523.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164495 AK149117.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179311 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10572618 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572617 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572621 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572619 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572620 - -
          Affymetrix HT MG-430 PM Array Plate 1429039_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1434281_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1429038_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10572616 - -
          Affymetrix Mouse Gene 1.1 ST Array 10579508 - -
          Mouse Genome 430 2.0 Array 1434281_PM_at - -
          Mouse Genome 430 2.0 Array 1429038_at - -
          Mouse Genome 430 2.0 Array 1429039_s_at - -
          Mouse Genome 430 2.0 Array 1434281_at - -
          Mouse Genome 430 2.0 Array 1429038_PM_at - -
          Mouse Genome 430 2.0 Array 1429039_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66498_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66498_at - -
          Affymetrix Mouse Clariom S Array PSR0800005695.mm.1 NM_025600.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005695.mm.1 TC0800000791.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800005697.mm.1 NM_025600.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005697.mm.1 TC0800000791.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800005694.mm.1 NM_025600.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005694.mm.1 TC0800000791.mm.1 MTA TC
          Mouse Expression Set 430 1434281_at - -
          Mouse Expression Set 430 1429038_at - -
          Mouse Expression Set 430 1429038_PM_at - -
          Mouse Expression Set 430 1429039_s_at - -
          Mouse Expression Set 430 1429039_PM_s_at - -
          Mouse Expression Set 430 1434281_PM_at - -
          Murine Genome U74v2 Set 102072_g_at - -
          Murine Genome U74v2 Set 102312_PM_r_at - -
          Murine Genome U74v2 Set 102071_PM_at - -
          Murine Genome U74v2 Set 97404_at - -
          Murine Genome U74v2 Set 102312_r_at - -
          Murine Genome U74v2 Set 102071_at - -
          Murine Genome U74v2 Set 161719_f_at - -
          Murine Genome U74v2 Set 161719_PM_f_at - -
          Murine Genome U74v2 Set 102072_PM_g_at - -
          Murine Genome U74v2 Set 97404_PM_at - -
          Mouse Genome 430A 2.0 Array 1434281_at - -
          Mouse Genome 430A 2.0 Array 1429039_s_at - -
          Mouse Genome 430A 2.0 Array 1429038_PM_at - -
          Mouse Genome 430A 2.0 Array 1429038_at - -
          Mouse Genome 430A 2.0 Array 1434281_PM_at - -
          Mouse Genome 430A 2.0 Array 1429039_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1429039_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1429039_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1429038_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1429038_at - -
          Affymetrix HT Mouse Genome 430A Array 1434281_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1434281_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 545521 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0