Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01342545_m1
          See other DLEC1 GE Assays ›
          Gene Symbol:
          Dlec1
          Gene Name:
          deleted in lung and esophageal cancer 1
          Gene Aliases:
          D630005C06Rik
          Chromosome Location:
          Chr.9: 119101535 - 119148246 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01342545_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dlec1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177117.3 NP_796091.2 2 - 3 603 60
          GenBank mRNA AK045848.1 - 2 - 3 604 60
          AK133987.1 - 2 - 3 574 60
          BC145277.1 - 2 - 3 586 60
          BC145278.1 - 2 - 3 586 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001572.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dlec1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2026114 NM_177117.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2026119 AK133987.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 448508 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 236646 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 19995 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 665155 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 730649 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 911434 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914568 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 938289 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 379814 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 591919 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 353320 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 835934 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 935832 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 129617 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 71877 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 102858 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 589934 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 66023 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 891168 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 925527 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 150087 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 707503 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 262976 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777071 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 43033 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 412925 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 922420 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 154604 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 69577 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 74161 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 66114 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 75014 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 64128 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127700 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168427 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127653 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2026114 NM_177117.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2026119 AK133987.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06945967 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127700 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02043854 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168427 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127653 - -
          Whole Mouse Genome G4122A A_52_P613788 AK045848.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115003 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168427 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127653 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08136509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07564647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07564656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08113358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08144917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07680437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07611811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07680439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07866524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07680442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07813675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08136506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07957425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07671631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07543426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07957415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07517592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07698925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07611818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26870612 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10590098 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590132 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590131 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590122 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590099 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590125 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590120 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590106 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590104 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590119 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590123 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590115 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590130 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590126 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590097 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590107 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590111 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590105 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590133 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590129 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590100 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590112 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590113 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590128 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590127 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590114 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590101 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590121 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590124 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590109 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590108 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590116 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590117 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590110 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590118 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590103 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590102 - -
          Affymetrix Mouse Gene 1.1 ST Array 10590096 - -
          Affymetrix Mouse Clariom S Array PSR0900013074.mm.1 NM_177117.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900013074.mm.1 NM_177117.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900013074.mm.1 TC0900001572.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 420986 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 7000180 - -
          Illumina mouse-6 v1.0 expression beadchip 7000180 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007000180 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2727768 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2727768 - -
          mouse-6 v1.1 (Illumina) scl0320256.5_3-S - -
          Illumina Sentrix mouse-6.v1 bead array 7000180 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0320256.5_3-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7000180 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2727768 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2727768 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline