Search
Search

Nr1i2| Gene Symbol: Nr1i2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098404.1 | NP_001091874.1 | 2 - 3 | 528 | 66 | |
| NM_010936.3 | NP_035066.1 | 2 - 3 | 528 | 66 | ||
| GenBank mRNA | AF031814.1 | - | 2 - 3 | 343 | 66 | |
| AI746915.1 | - | 2 - 3 | 528 | 66 | ||
| AK011108.1 | - | 2 - 3 | 497 | 66 | ||
| AK018630.1 | - | 2 - 3 | 528 | 66 | ||
| BC120796.1 | - | 2 - 3 | 275 | 40129719 | 66 | |
| BC137780.1 | - | 2 - 3 | 275 | 8860799 | 66 | |
Set Membership: |
|