Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01344531_m1
          See other RWDD2B GE Assays ›
          Gene Symbol:
          Rwdd2b
          Gene Name:
          RWD domain containing 2B
          Gene Aliases:
          ORF5
          Chromosome Location:
          Chr.16: 87433331 - 87440592 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01344531_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rwdd2b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016924.2 NP_058620.2 4 - 5 802 89
          GenBank mRNA AF177771.1 - 4 - 5 758 89
          AK002746.1 - 4 - 5 768 89
          AK084966.1 - 4 - 5 777 89
          BC002143.1 - 4 - 5 783 3484719 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600001985.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rwdd2b

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30939717 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30939754 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30939712 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30939716 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30939711 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30939756 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30939755 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30939715 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30939710 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371255 AK002746.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371255 AK084966.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371255 BC002143.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371255 NM_016924.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371255 AF177771.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371255 AK002746.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371255 AK084966.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371255 BC002143.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371255 NM_016924.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P371255 AF177771.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 282292 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 782518 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 449824 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 903701 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 493616 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 110477 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131002 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168632 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147127 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371255 AK002746.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371255 AK084966.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371255 BC002143.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371255 NM_016924.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371255 AF177771.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371255 AK002746.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371255 AK084966.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371255 BC002143.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371255 NM_016924.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P371255 AF177771.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939759 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147127 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168632 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131002 - -
          Whole Mouse Genome G4122A A_51_P371255 AK002746.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371255 AK084966.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371255 BC002143.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371255 NM_016924.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371255 AF177771.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371255 AK002746.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371255 AK084966.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371255 BC002143.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371255 NM_016924.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371255 AF177771.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136750 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131002 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147127 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168632 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12556634 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12556633 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12729435 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12754650 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12556636 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12556640 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30939716 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30939712 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30939710 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30939715 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30939755 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30939711 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30939754 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30939717 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30939756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939735 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12819787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12809170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12565003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12609152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939759 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939782 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939748 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13014197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939779 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12791809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939756 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12839012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12564999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939742 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939749 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12565008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939745 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939766 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939763 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939734 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30939736 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10440595 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440596 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440597 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440599 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440594 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440598 - -
          Affymetrix HT MG-430 PM Array Plate 1419760_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10440593 - -
          Mouse Genome 430 2.0 Array 1419760_PM_a_at - -
          Mouse Genome 430 2.0 Array 1419760_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 53858_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 53858_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1600015407.mm.1 NM_016924.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015407.mm.1 NM_016924.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015407.mm.1 TC1600001985.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600015408.mm.1 NM_016924.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015408.mm.1 NM_016924.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015408.mm.1 TC1600001985.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600015409.mm.1 NM_016924.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015409.mm.1 NM_016924.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015409.mm.1 TC1600001985.mm.1 MTA TC
          Mouse Expression Set 430 1419760_PM_a_at - -
          Mouse Expression Set 430 1419760_a_at - -
          Mouse Genome 430A 2.0 Array 1419760_a_at - -
          Mouse Genome 430A 2.0 Array 1419760_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1419760_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1419760_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 757868 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 770465 - -
          Illumina mouse-6 v1.0 expression beadchip 770465 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000770465 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1239330 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1239330 - -
          mouse-6 v1.1 (Illumina) scl48262.5_262-S - -
          Illumina Sentrix mouse-6.v1 bead array 770465 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl48262.5_262-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 770465 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1239330 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1239330 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0