Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01344699_g1
          See other CSTA1 GE Assays ›
          Gene Symbol:
          Csta1
          Gene Name:
          cystatin A1
          Gene Aliases:
          Csta
          Chromosome Location:
          Chr.16: 36119946 - 36131189 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01344699_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Csta1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033239.3 NP_001028411.1 1 - 2 207 115
          GenBank mRNA AK137702.1 - 1 - 2 208 115
          BC139156.1 - 1 - 2 102 8862178 115
          BC139157.1 - 1 - 2 102 8862179 115

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of peptidase activity
          negative regulation of endopeptidase activity
          protease binding
          endopeptidase inhibitor activity
          cysteine-type endopeptidase inhibitor activity
          structural molecule activity
          peptidase inhibitor activity
          protein binding, bridging

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2069724 NM_001033239.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2069724 AK137702.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916347 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 690323 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 770541 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 891877 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 933995 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 520022 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 540957 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 39813 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 82292 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2069724 NM_001033239.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2069724 AK137702.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30673882 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03170185 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03170174 - -
          Whole Mouse Genome G4122A A_52_P686716 BC139157.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P686716 NM_001033239.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P686716 AK137702.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P686716 BC139156.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03170185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30673834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12774043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12829330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12576396 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12760922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13008827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12819956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12819955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12593414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12761554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12904454 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12812979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30673846 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10439283 - -
          Affymetrix Mouse Gene 1.0 ST Array 10439284 - -
          Affymetrix HT MG-430 PM Array Plate 1435760_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10439282 - -
          Mouse Genome 430 2.0 Array 1435760_PM_at - -
          Mouse Genome 430 2.0 Array 1435760_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 209294_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 209294_PM_at - -
          Mouse Expression Set 430 1435760_at - -
          Mouse Expression Set 430 1435760_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435760_at - -
          Affymetrix HT Mouse Genome 430B Array 1435760_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2945358 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2945358 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline