Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01348938_m1
          See other CTS6 GE Assays ›
          Gene Symbol:
          Cts6
          Gene Name:
          cathepsin 6
          Gene Aliases:
          1600022N02Rik
          Chromosome Location:
          Chr.13: 61195147 - 61203392 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01348938_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cts6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021445.1 NP_067420.1 6 - 7 849 100
          GenBank mRNA AF223401.1 - 6 - 7 849 100
          AK005518.1 - 6 - 7 868 100
          AY014781.1 - 6 - 7 865 100
          BC099455.1 - 6 - 7 851 30787134 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002148.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protease 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cts6

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          proteolysis involved in cellular protein catabolic process
          molecular_function
          cysteine-type endopeptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P229893 AY014781.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P229893 AF223401.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P229893 AK005518.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P229893 BC099455.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P229893 NM_021445.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P229893 AY014781.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P229893 AF223401.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P229893 AK005518.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P229893 BC099455.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P229893 NM_021445.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 68028 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 300580 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358773 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 935077 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 704544 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 93775 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161553 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P229893 AY014781.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P229893 AF223401.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P229893 AK005518.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P229893 BC099455.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P229893 NM_021445.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P229893 AY014781.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P229893 AF223401.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P229893 AK005518.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P229893 BC099455.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P229893 NM_021445.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P229893 AY014781.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P229893 AF223401.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P229893 AK005518.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P229893 BC099455.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P229893 NM_021445.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P229893 AY014781.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P229893 AF223401.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P229893 AK005518.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P229893 BC099455.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P229893 NM_021445.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29088916 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161553 - -
          Whole Mouse Genome G4122A A_51_P229893 AY014781.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P229893 AF223401.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P229893 AK005518.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P229893 BC099455.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P229893 NM_021445.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P229893 AY014781.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P229893 AF223401.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P229893 AK005518.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P229893 BC099455.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P229893 NM_021445.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129306 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161553 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10982060 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10982068 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10982054 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10982045 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10982052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29088938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10906093 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10725852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10982060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10982049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10906102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10982048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10725850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29088931 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10409917 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409923 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409918 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409922 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409920 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409919 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409921 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409916 - -
          Affymetrix HT MG-430 PM Array Plate 1418689_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10409915 - -
          Mouse Genome 430 2.0 Array 1418689_at - -
          Mouse Genome 430 2.0 Array 1418689_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 58518_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 58518_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300013806.mm.1 NM_021445.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013806.mm.1 NM_021445.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013806.mm.1 TC1300002148.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013807.mm.1 NM_021445.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013807.mm.1 NM_021445.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013807.mm.1 TC1300002148.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013808.mm.1 NM_021445.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013808.mm.1 NM_021445.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013808.mm.1 TC1300002148.mm.1 MTA TC
          Mouse Expression Set 430 1418689_at - -
          Mouse Expression Set 430 1418689_PM_at - -
          Mouse Genome 430A 2.0 Array 1418689_at - -
          Mouse Genome 430A 2.0 Array 1418689_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418689_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418689_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 879959 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2630184 - -
          Illumina mouse-6 v1.0 expression beadchip 2630184 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002630184 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2665568 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2665568 - -
          mouse-6 v1.1 (Illumina) scl43858.8.4_9-S - -
          Illumina Sentrix mouse-6.v1 bead array 2630184 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl43858.8.4_9-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2630184 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2665568 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2665568 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline