Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01351404_g1
          See other PIGO GE Assays ›
          Gene Symbol:
          Pigo
          Gene Name:
          phosphatidylinositol glycan anchor biosynthesis, class O
          Gene Aliases:
          PIG-O
          Chromosome Location:
          Chr.4: 43017637 - 43027696 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01351404_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pigo
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AB038560.1 - 9 - 10 3835 67
          AK013913.1 - 3 - 4 386 67
          AK050819.1 - 10 - 11 3176 67
          AK220315.1 - 9 - 10 3783 67
          BC025857.1 - 4 - 5 1316 67
          BC036977.1 - 10 - 11 3108 67
          BC092394.1 - 9 - 10 3853 30534262 67
          BC141058.1 - 9 - 10 3791 9055670 67
          BU708685.1 - 1 - 2 83 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400002515.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2042202 BC092394.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2042202 AK013913.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2042202 BC025857.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2042202 NM_020035.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2042202 AB038560.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 73435 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 759926 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 214606 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 812174 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755482 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 278643 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 24864 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174980 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2042202 BC092394.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2042202 AK013913.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2042202 BC025857.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2042202 NM_020035.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2042202 AB038560.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P462081 BC036977.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P462081 BC092394.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P462081 BC141058.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P462081 AK050819.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P462081 AK013913.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22869571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22869564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22869553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22869567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22869562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22869574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22869550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22869558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22869563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22869549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22869570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22869559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22869555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22869561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22869554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22869566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22869569 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05216028 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00857646 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174980 - -
          Whole Mouse Genome G4122A A_52_P462081 BC036977.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P462081 BC092394.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P462081 BC141058.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P462081 AK050819.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P462081 AK013913.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113323 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174980 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03569178 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03569174 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03768750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22869511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04010298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03569175 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22869520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22869513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03597022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03597008 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22869558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22869551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22869559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03270689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22869562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22869505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22869567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03597023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22869542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22869556 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03597011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03597004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03596999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03524651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22869547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03569168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03597010 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22869531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22869571 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22869572 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22869566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03569170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22869545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22869553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22869511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03409687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22869522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22869549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22869555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03597027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22869516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22869573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03597012 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10512440 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512437 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512442 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512438 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512435 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512432 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512433 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512439 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512434 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512441 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512436 - -
          Affymetrix HT MG-430 PM Array Plate 1438155_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1415992_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1437142_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10512431 - -
          Mouse Genome 430 2.0 Array 1438155_x_at - -
          Mouse Genome 430 2.0 Array 1438155_PM_x_at - -
          Mouse Genome 430 2.0 Array 1415992_at - -
          Mouse Genome 430 2.0 Array 1437142_a_at - -
          Mouse Genome 430 2.0 Array 1437142_PM_a_at - -
          Mouse Genome 430 2.0 Array 1415992_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56703_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56703_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400020351.mm.1 TC0400002515.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020351.mm.1 NM_020035.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020353.mm.1 TC0400002515.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020353.mm.1 NM_020035.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020352.mm.1 TC0400002515.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020352.mm.1 NM_020035.3 Refseq
          Mouse Expression Set 430 1415992_PM_at - -
          Mouse Expression Set 430 1437142_a_at - -
          Mouse Expression Set 430 1438155_x_at - -
          Mouse Expression Set 430 1438155_PM_x_at - -
          Mouse Expression Set 430 1437142_PM_a_at - -
          Mouse Expression Set 430 1415992_at - -
          Murine Genome U74v2 Set 168756_at - -
          Murine Genome U74v2 Set 168756_PM_at - -
          Mouse Genome 430A 2.0 Array 1415992_PM_at - -
          Mouse Genome 430A 2.0 Array 1415992_at - -
          Mouse Genome 430A 2.0 Array 1437142_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1437142_a_at - -
          Mouse Genome 430A 2.0 Array 1438155_x_at - -
          Mouse Genome 430A 2.0 Array 1438155_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1415992_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1438155_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1437142_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1415992_at - -
          Affymetrix HT Mouse Genome 430A Array 1437142_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1438155_x_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2340026 - -
          MouseWG-6 v2 Expression BeadChip 5340097 - -
          Illumina mouse-6 v1.0 expression beadchip 1400528 - -
          Illumina mouse-6 v1.0 expression beadchip 2340026 - -
          Illumina mouse-6 v1.0 expression beadchip 5340097 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002340026 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005340097 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001400528 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1244529 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2774001 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2740372 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2774001 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2740372 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244529 - -
          mouse-6 v1.1 (Illumina) scl24409.10.1_30-S - -
          mouse-6 v1.1 (Illumina) scl0002789.1_1177-S - -
          mouse-6 v1.1 (Illumina) scl0002634.1_125-S - -
          Illumina Sentrix mouse-6.v1 bead array 5340097 - -
          Illumina Sentrix mouse-6.v1 bead array 1400528 - -
          Illumina Sentrix mouse-6.v1 bead array 2340026 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0002634.1_125-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0002789.1_1177-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24409.10.1_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2340026 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5340097 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1400528 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1229551 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2997002 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2740372 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2774001 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2997002 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2740372 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1229551 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl24409.10.1_30-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline