Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01351549_g1
          See other FAM166B GE Assays ›
          Gene Symbol:
          Fam166b
          Gene Name:
          family with sequence similarity 166, member B
          Gene Aliases:
          4833436C18Rik
          Chromosome Location:
          Chr.4: 43426675 - 43431593 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01351549_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam166b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001162381.1 NP_001155853.1 2 - 3 323 62
          NM_177377.5 NP_796351.2 2 - 3 323 62
          GenBank mRNA AK053208.1 - 2 - 3 324 62
          BC172030.1 - 2 - 3 306 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400002522.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fam166b

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P137660 NM_001162381.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P137660 NM_177377.5 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P137660 AK029435.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P137660 AK053208.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048460 NM_001162381.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048460 NM_177377.5 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048460 AK029435.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048460 AK053208.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048460 BC172030.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 470399 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120689 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P137660 NM_001162381.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P137660 NM_177377.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P137660 AK029435.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P137660 AK053208.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048460 NM_001162381.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048460 NM_177377.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048460 AK029435.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048460 AK053208.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048460 BC172030.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871697 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120689 - -
          Whole Mouse Genome G4122A A_52_P199237 NM_001162381.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P199237 NM_177377.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P199237 AK029435.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P199237 AK053208.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P199237 BC172030.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137660 NM_001162381.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137660 NM_177377.5 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137660 AK029435.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137660 AK053208.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120689 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03557551 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03557557 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03923377 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03557549 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03923374 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03557555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03926645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03756179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03584916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871657 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03272393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03557558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03893186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03562238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03663848 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03931130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03756178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03756174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03557551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03923374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871651 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10512467 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512466 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512468 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512469 - -
          Affymetrix Mouse Gene 1.1 ST Array 10512465 - -
          Affymetrix Mouse Clariom S Array PSR0400020477.mm.1 TC0400002522.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020477.mm.1 NM_001162381.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020477.mm.1 NM_001162381.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020477.mm.1 NM_177377.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020477.mm.1 NM_177377.5 Refseq
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3990722 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003990722 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2692952 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2692952 - -
          mouse-6 v1.1 (Illumina) scl0329831.1_281-S - -
          Illumina Sentrix mouse-6.v1 bead array 3990722 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0329831.1_281-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3990722 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2692952 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2692952 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0329831.1_281-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline