Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01613328_g1
          See other PSAT1 GE Assays ›
          Gene Symbol:
          Psat1
          Gene Name:
          phosphoserine aminotransferase 1
          Gene Aliases:
          D8Ertd814e, EPIP, PSA, Psat
          Chromosome Location:
          Chr.19: 15905123 - 15925059 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01613328_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psat1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177420.2 NP_803155.1 2 - 3 253 165
          GenBank mRNA AF259674.1 - 2 - 3 151 165
          AK032487.1 - 2 - 3 254 165
          AK035361.1 - 2 - 3 143 165
          AK049855.1 - 2 - 3 187 165
          AK090108.1 - 2 - 3 181 165
          AK145672.1 - 2 - 3 178 165
          AK162609.1 - 2 - 3 144 165
          AK166880.1 - 2 - 3 178 165
          AK167029.1 - 2 - 3 144 165
          AK167044.1 - 2 - 3 191 165
          AK168098.1 - 2 - 3 168 165
          AK168216.1 - 2 - 3 191 165
          AK169370.1 - 2 - 3 168 165
          BC004827.1 - 2 - 3 219 2616298 165

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AF259674.1, AK049855.1, AK090108.1, AK145672.1, AK162609.1, AK166880.1, AK167029.1, AK167044.1, AK168098.1, AK169370.1, BC004827.1, NM_177420.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psat1

          Panther Classification:

          Molecular Function -

          transaminase

          Gene Ontology Categories:

          Function(s) Process(es)

          L-serine biosynthetic process
          metabolic process
          cellular amino acid biosynthetic process
          catalytic activity
          O-phospho-L-serine:2-oxoglutarate aminotransferase activity
          transaminase activity
          transferase activity
          pyridoxal phosphate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052290 BC004827.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052290 BC005657.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052290 AK169370.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052290 AK168098.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052290 AK168216.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P121495 BC004827.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P121495 AK169370.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P121495 AK167044.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P121495 AK168098.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P121495 AK168216.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 867091 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 702194 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 823255 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 688487 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 322555 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732099 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 920308 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 155947 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 60499 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 363702 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 17240 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 184877 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 627465 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 127957 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 9804 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158531 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052290 BC004827.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052290 BC005657.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052290 AK169370.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052290 AK168098.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052290 AK168216.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P121495 BC004827.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P121495 AK169370.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P121495 AK167044.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P121495 AK168098.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P121495 AK168216.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P317427 BC004827.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P317427 BC005657.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P317427 AK169370.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P317427 AK191308.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P317427 AK167044.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31977240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31977226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31977228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31977236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31977232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31977237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31977233 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03556586 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158531 - -
          Whole Mouse Genome G4122A A_51_P317427 BC004827.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317427 BC005657.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317427 AK169370.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317427 AK191308.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317427 AK167044.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158531 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14281082 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14281086 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14210587 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14281083 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14210593 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14281080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31977085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14371495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977226 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977076 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14371496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14142501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14371503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14232065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14232039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977248 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977245 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31977089 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10466415 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466418 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466417 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466412 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466411 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466416 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466413 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466419 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466421 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466420 - -
          Affymetrix HT MG-430 PM Array Plate 1451064_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1454607_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10466410 - -
          Mouse Genome 430 2.0 Array 1451064_PM_a_at - -
          Mouse Genome 430 2.0 Array 1451064_a_at - -
          Mouse Genome 430 2.0 Array 1454607_s_at - -
          Mouse Genome 430 2.0 Array 1454607_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 107272_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 107272_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900010949.mm.1 NM_001205339.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010949.mm.1 NM_001205339.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010949.mm.1 NM_177420.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010949.mm.1 NM_177420.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010949.mm.1 TC1900001203.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010939.mm.1 NM_001205339.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010939.mm.1 NM_001205339.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010939.mm.1 NM_177420.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010939.mm.1 NM_177420.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010939.mm.1 TC1900001203.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010941.mm.1 NM_001205339.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010941.mm.1 NM_001205339.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010941.mm.1 NM_177420.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010941.mm.1 NM_177420.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010941.mm.1 TC1900001203.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010940.mm.1 NM_001205339.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010940.mm.1 NM_001205339.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010940.mm.1 NM_177420.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010940.mm.1 NM_177420.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010940.mm.1 TC1900001203.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010947.mm.1 NM_001205339.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010947.mm.1 NM_001205339.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010947.mm.1 NM_177420.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010947.mm.1 NM_177420.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010947.mm.1 TC1900001203.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010948.mm.1 NM_001205339.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010948.mm.1 NM_001205339.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010948.mm.1 NM_177420.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010948.mm.1 NM_177420.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010948.mm.1 TC1900001203.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010951.mm.1 NM_001205339.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010951.mm.1 NM_001205339.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010951.mm.1 NM_177420.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010951.mm.1 NM_177420.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010951.mm.1 TC1900001203.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010943.mm.1 NM_001205339.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010943.mm.1 NM_001205339.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010943.mm.1 NM_177420.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010943.mm.1 NM_177420.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010943.mm.1 TC1900001203.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010944.mm.1 NM_001205339.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010944.mm.1 NM_001205339.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010944.mm.1 NM_177420.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010944.mm.1 NM_177420.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010944.mm.1 TC1900001203.mm.1 MTA TC
          Mouse Expression Set 430 1454607_s_at - -
          Mouse Expression Set 430 1454607_PM_s_at - -
          Murine Genome U74v2 Set 96295_at - -
          Murine Genome U74v2 Set 96295_PM_at - -
          Mouse Genome 430A 2.0 Array 1454607_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1454607_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1451064_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1451064_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1454607_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1454607_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 893894 - -
          ABI Mouse Genome Survey Microarray 916661 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 670446 - -
          MouseWG-6 v2 Expression BeadChip 1570102 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1221048 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2598243 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221048 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2598243 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1221048 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline