Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01615849_mH
          See other SULT2A4 GE Assays ›
          Gene Symbol:
          Sult2a4
          Gene Name:
          sulfotransferase family 2A, dehydroepiandrosterone (DHEA)-preferring, member 4
          Gene Aliases:
          EG434121, Gm5584
          Chromosome Location:
          Chr.7: 13909677 - 13989591 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01615849_mH
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sult2a4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001101534.1 NP_001095004.1 5 - 6 745 111

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002373.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          N-acetylglucosamine 6-O-sulfotransferase activity
          N-acetylgalactosamine 4-O-sulfotransferase activity
          alcohol sulfotransferase activity
          heparan sulfate 2-O-sulfotransferase activity
          drug binding
          sulfotransferase activity
          HNK-1 sulfotransferase activity
          transferase activity
          heparan sulfate 6-O-sulfotransferase activity
          trans-9R,10R-dihydrodiolphenanthrene sulfotransferase activity
          1-phenanthrol sulfotransferase activity
          3-phenanthrol sulfotransferase activity
          4-phenanthrol sulfotransferase activity
          trans-3,4-dihydrodiolphenanthrene sulfotransferase activity
          9-phenanthrol sulfotransferase activity
          2-phenanthrol sulfotransferase activity
          phenanthrol sulfotransferase activity
          1-hydroxypyrene sulfotransferase activity
          3'-phosphoadenosine 5'-phosphosulfate binding
          galactose 3-O-sulfotransferase activity
          proteoglycan sulfotransferase activity
          cholesterol sulfotransferase activity
          hydroxyjasmonate sulfotransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952507 NM_001101534.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P218833 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 319062 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 794151 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 607376 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 960223 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 611942 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 125310 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 35032 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 172708 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952507 NM_001101534.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24976717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24976785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24976694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24976754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24976695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24976719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24976643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24976809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24976698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24976783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24976716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24976656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24976659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24976725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24976787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24976697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24976787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24976785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06496235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06496244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24976809 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10560112 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560114 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560113 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560116 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560115 - -
          Affymetrix Mouse Gene 1.1 ST Array 10560111 - -
          Affymetrix Mouse Clariom S Array PSR0700021042.mm.1 TC0700002373.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021042.mm.1 NM_001101534.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021042.mm.1 NM_001101534.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021043.mm.1 TC0700002373.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021043.mm.1 NM_001101534.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021043.mm.1 NM_001101534.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021045.mm.1 TC0700002373.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021045.mm.1 NM_001101534.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021045.mm.1 NM_001101534.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021041.mm.1 TC0700002373.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021041.mm.1 NM_001101534.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021041.mm.1 NM_001101534.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021044.mm.1 TC0700002373.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021044.mm.1 NM_001101534.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021044.mm.1 NM_001101534.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021046.mm.1 TC0700002373.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021046.mm.1 NM_001101534.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021046.mm.1 NM_001101534.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 805316 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline