Search
Search
| Gene Symbol: Otx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011023.3 | NP_035153.1 | 4 - 5 | 526 | 127 | |
| GenBank mRNA | AK042975.1 | - | 4 - 5 | 490 | 127 | |
| AK050758.1 | - | 4 - 5 | 487 | 127 | ||
| AK083434.1 | - | 4 - 5 | 488 | 127 | ||
| AK083671.1 | - | 4 - 5 | 470 | 127 | ||
| AK158588.1 | - | 2 - 3 | 2884 | 127 | ||
| BC057105.1 | - | 3 - 4 | 436 | 6416436 | 127 | |
| BC058354.1 | - | 3 - 4 | 394 | 6816961 | 127 | |
Set Membership: |
|