Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01700149_s1
          See other TAS2R144 GE Assays ›
          Gene Symbol:
          Tas2r144
          Gene Name:
          taste receptor, type 2, member 144
          Gene Aliases:
          Tas2r40, Tas2r44, mt2r33
          Chromosome Location:
          Chr.6: 42215328 - 42216287 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01700149_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tas2r144
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001001453.1 NP_001001453.1 1 866 83

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tas2r144

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of chemical stimulus involved in sensory perception of bitter taste
          signal transduction
          G-protein coupled receptor signaling pathway
          response to stimulus
          sensory perception of taste
          signal transducer activity
          G-protein coupled receptor activity
          taste receptor activity
          bitter taste receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004646 NM_001001453.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170835 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004646 NM_001001453.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396927 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170835 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136486 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170835 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05372860 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05372856 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05372859 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05372855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05372853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05381864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05774019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396949 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10537711 - -
          Affymetrix Mouse Gene 1.1 ST Array 10537710 - -
          Affymetrix Mouse Clariom S Array PSR0600003600.mm.1 TC0600000453.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600003600.mm.1 NM_001001453.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600003600.mm.1 NM_001001453.1 Refseq
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160931 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160931 - -
          mouse-6 v1.1 (Illumina) GI_47824892-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160931 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160931 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0