Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01716144_m1
          See other IGLL1 GE Assays ›
          Gene Symbol:
          Igll1
          Gene Name:
          immunoglobulin lambda-like polypeptide 1
          Gene Aliases:
          BB139905, Igl-5, Igll, Lambda5
          Chromosome Location:
          Chr.16: 16860671 - 16863985 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01716144_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Igll1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001190325.1 NP_001177254.1 2 - 3 409 64
          GenBank mRNA M30387.1 - 2 - 3 238 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600001270.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          immunoglobulin

          Gene Ontology Categories:

          Process(es)

          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P493519 AK137552.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P493519 NM_001190325.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P493519 AK137514.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P493519 AK137552.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P493519 NM_001190325.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P493519 AK137514.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159563 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P493519 AK137552.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P493519 NM_001190325.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P493519 AK137514.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P493519 AK137552.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P493519 NM_001190325.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P493519 AK137514.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159563 - -
          Whole Mouse Genome G4122A A_52_P493519 AK137552.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P493519 NM_001190325.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P493519 AK137514.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P493519 AK137552.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P493519 NM_001190325.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P493519 AK137514.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P273394 AK137552.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P273394 AK137514.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13037742 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12943969 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10438061 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438063 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438062 - -
          Affymetrix Mouse Gene 1.1 ST Array 10438060 - -
          Affymetrix Mouse Clariom S Array PSR1600010283.mm.1 NM_001190325.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010283.mm.1 NM_001190325.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010283.mm.1 TC1600001270.mm.1 MTA TC
          Murine Genome U74v2 Set 99429_PM_at - -
          Murine Genome U74v2 Set 99429_at - -
          Murine Genome U74v2 Set 99428_at - -
          Murine Genome U74v2 Set 99428_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 785449 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline