Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01716261_m1
          See other 9530036M11RIK GE Assays ›
          Gene Symbol:
          9530036M11Rik
          Gene Name:
          RIKEN cDNA 9530036M11 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 54139523 - 54156068 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01716261_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9530036M11Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK020588.1 - 2 - 3 860 133
          AK137178.1 - 1 - 2 241 133

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2225178 AK137178.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2225178 AK137178.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05739417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05634087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05380517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05077680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05634101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05380507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05739425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05077689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05077686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05828332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05739420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05077684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05380515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05634086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05634094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05077685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05380518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05380506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05380520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05077688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05739421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05739412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24461501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05739404 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1454146_PM_at - -
          Mouse Genome 430 2.0 Array 1454146_at - -
          Mouse Genome 430 2.0 Array 1454146_PM_at - -
          Mouse Expression Set 430 1454146_at - -
          Mouse Expression Set 430 1454146_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454146_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454146_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 596293 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103990632 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline