Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01717650_s1
          See other PHTF1OS GE Assays ›
          Gene Symbol:
          Phtf1os
          Gene Name:
          putative homeodomain transcription factor 1, opposite strand
          Gene Aliases:
          A130049A11Rik
          Chromosome Location:
          Chr.3: 103964536 - 103968316 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01717650_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Phtf1os
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_030676.1 1 1346 130
          GenBank mRNA AK037774.1 - 1 1326 130
          AK048683.1 - 1 1347 130
          AK051503.1 - 1 1344 130

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2214408 AK048683.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109455 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2214408 AK048683.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P299561 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109455 - -
          Whole Mouse Genome G4122A A_52_P299561 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109455 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10500846 - -
          Affymetrix Mouse Gene 1.1 ST Array 10500845 - -
          Affymetrix Mouse Clariom S Array PSR0300020594.mm.1 NR_030676.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020594.mm.1 NR_030676.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020594.mm.1 TC0300002635.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300020593.mm.1 NR_030676.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020593.mm.1 NR_030676.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020593.mm.1 TC0300002635.mm.1 MTA TC
          Murine Genome U74v2 Set 170671_PM_at - -
          Murine Genome U74v2 Set 170671_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 334282 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101570647 - -
          MouseWG-6 v2 Expression BeadChip 101340075 - -
          Illumina mouse-6 v1.0 expression beadchip 101340075 - -
          Illumina Sentrix mouse-6.v1 bead array 101570647 - -
          Illumina Sentrix mouse-6.v1 bead array 101340075 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101570647 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101340075 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1250256 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2573168 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0