Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01964671_s1
          See other INSL6 GE Assays ›
          Gene Symbol:
          Insl6
          Gene Name:
          insulin-like 6
          Gene Aliases:
          RIF1
          Chromosome Location:
          Chr.19: 29321354 - 29325318 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01964671_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Insl6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013754.1 NP_038782.1 2 439 130
          GenBank mRNA AF143807.2 - 2 439 130
          AK005622.1 - 2 476 130
          BC028752.1 - 2 451 1329516 130

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Insl6

          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          spermatogenesis
          spermatid development
          male gonad development
          fertilization
          sperm motility
          negative regulation of apoptotic process
          hormone activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459108 NM_013754.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459108 AF143807.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459108 AK005622.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459108 BC028752.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459108 NM_013754.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459108 AF143807.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459108 AK005622.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459108 BC028752.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 604301 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 137702 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 607752 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 587037 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 2025 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152833 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459108 NM_013754.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459108 AF143807.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459108 AK005622.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459108 BC028752.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459108 NM_013754.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459108 AF143807.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459108 AK005622.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459108 BC028752.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P459108 NM_013754.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P459108 AF143807.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P459108 AK005622.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P459108 BC028752.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P459108 NM_013754.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P459108 AF143807.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P459108 AK005622.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P459108 BC028752.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32052265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32052256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32052270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32052281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32052258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32052290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32052272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32052271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32052263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32052273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32052279 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32052291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32052288 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32052260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32052269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32052268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32052284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32052283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32052277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32052285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32052278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32052275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32052264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32052287 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32052274 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152833 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09146457 - -
          Whole Mouse Genome G4122A A_51_P459108 NM_013754.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459108 AF143807.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459108 AK005622.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459108 BC028752.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459108 NM_013754.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459108 AF143807.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459108 AK005622.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459108 BC028752.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107704 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152833 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14206134 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14479254 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14297291 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14206137 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14297289 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14479253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32052297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052291 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052273 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052253 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052256 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14484179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052295 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14484183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052292 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052271 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052279 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052269 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14206137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052288 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052283 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14406795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052287 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052280 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052277 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052272 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052245 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052248 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14206152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052263 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32052251 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10466933 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466934 - -
          Affymetrix HT MG-430 PM Array Plate 1418346_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10466932 - -
          Mouse Genome 430 2.0 Array 1418346_PM_at - -
          Mouse Genome 430 2.0 Array 1418346_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 27356_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 27356_at - -
          Affymetrix Mouse Clariom S Array PSR1900011884.mm.1 NM_013754.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900011884.mm.1 NM_013754.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900011884.mm.1 TC1900001340.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900011885.mm.1 NM_013754.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900011885.mm.1 NM_013754.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900011885.mm.1 TC1900001340.mm.1 MTA TC
          Mouse Expression Set 430 1418346_at - -
          Mouse Expression Set 430 1418346_PM_at - -
          Mouse Genome 430A 2.0 Array 1418346_at - -
          Mouse Genome 430A 2.0 Array 1418346_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418346_at - -
          Affymetrix HT Mouse Genome 430A Array 1418346_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 924952 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5570497 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005570497 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2677859 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2677859 - -
          mouse-6 v1.1 (Illumina) scl52596.2.1_112-S - -
          Illumina Sentrix mouse-6.v1 bead array 5570497 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52596.2.1_112-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5570497 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2677859 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2677859 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline