Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01972246_s1
          See other UBD GE Assays ›
          Gene Symbol:
          Ubd
          Gene Name:
          ubiquitin D
          Gene Aliases:
          FAT10
          Chromosome Location:
          Chr.17: 37193892 - 37196101 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01972246_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ubd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023137.3 NP_075626.1 2 719 170
          GenBank mRNA AK008552.1 - 2 692 170

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ubd

          Gene Ontology Categories:

          Function(s) Process(es)

          ubiquitin-dependent protein catabolic process
          protein ubiquitination
          protein modification by small protein conjugation
          myeloid dendritic cell differentiation
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          aggresome assembly
          protein binding
          proteasome binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338066 NM_023137.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338066 AK008116.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338066 AK008552.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338066 BC027627.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338066 BC036383.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338066 NM_023137.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338066 AK008116.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338066 AK008552.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338066 BC027627.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338066 BC036383.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 884762 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137392 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338066 NM_023137.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338066 AK008116.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338066 AK008552.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338066 BC027627.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338066 BC036383.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338066 NM_023137.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338066 AK008116.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338066 AK008552.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338066 BC027627.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338066 BC036383.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167725 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167758 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137392 - -
          Whole Mouse Genome G4122A A_52_P338066 NM_023137.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338066 AK008116.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338066 AK008552.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338066 BC027627.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338066 BC036383.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338066 NM_023137.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338066 AK008116.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338066 AK008552.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338066 BC027627.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338066 BC036383.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123759 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137392 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13553348 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13553343 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13240603 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13240601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13179812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13179822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13062873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13248952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167747 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13553345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167772 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13553344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13255108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13179823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167766 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167734 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13553348 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167724 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167769 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167742 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167763 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13240604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167773 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10445116 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445113 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445115 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445114 - -
          Affymetrix HT MG-430 PM Array Plate 1419762_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10445112 - -
          Mouse Genome 430 2.0 Array 1419762_at - -
          Mouse Genome 430 2.0 Array 1419762_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 24108_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 24108_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700007674.mm.1 NM_023137.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007674.mm.1 NM_023137.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007674.mm.1 TC1700000761.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007677.mm.1 NM_023137.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007677.mm.1 NM_023137.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007677.mm.1 TC1700000761.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007676.mm.1 NM_023137.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007676.mm.1 NM_023137.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007676.mm.1 TC1700000761.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007675.mm.1 NM_023137.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007675.mm.1 NM_023137.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007675.mm.1 TC1700000761.mm.1 MTA TC
          Mouse Expression Set 430 1419762_at - -
          Mouse Expression Set 430 1419762_PM_at - -
          Murine Genome U74v2 Set 92715_PM_at - -
          Murine Genome U74v2 Set 92715_at - -
          Murine Genome U74v2 Set 168938_PM_r_at - -
          Murine Genome U74v2 Set 169362_f_at - -
          Murine Genome U74v2 Set 168938_r_at - -
          Murine Genome U74v2 Set 169362_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1419762_at - -
          Mouse Genome 430A 2.0 Array 1419762_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419762_at - -
          Affymetrix HT Mouse Genome 430A Array 1419762_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 920469 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5570632 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005570632 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2426853 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2426853 - -
          mouse-6 v1.1 (Illumina) scl50736.3.1_4-S - -
          Illumina Sentrix mouse-6.v1 bead array 5570632 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50736.3.1_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5570632 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2426853 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2426853 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline