Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01972453_s1
          See other CYP2B10 GE Assays ›
          Gene Symbol:
          Cyp2b10
          Gene Name:
          cytochrome P450, family 2, subfamily b, polypeptide 10
          Gene Aliases:
          Cyp2b, Cyp2b20, p16
          Chromosome Location:
          Chr.7: 25897658 - 25926624 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01972453_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp2b10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009999.4 NP_034129.1 9 1745 120
          GenBank mRNA AF128849.1 - 9 1727 120
          AF129405.1 - 8 1541 120
          AK028103.1 - 9 1745 120
          BC060973.1 - 9 1739 30301385 120

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyp2b10

          Panther Classification:

          Molecular Function -

          oxidoreductase oxygenase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          xenobiotic metabolic process
          steroid metabolic process
          drug metabolic process
          epoxygenase P450 pathway
          cellular ketone metabolic process
          exogenous drug catabolic process
          oxidation-reduction process
          monooxygenase activity
          iron ion binding
          arachidonic acid epoxygenase activity
          steroid hydroxylase activity
          oxidoreductase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
          oxygen binding
          heme binding
          metal ion binding
          aromatase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2044653 AK028103.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2044653 BC060973.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2044653 NM_009999.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 795716 AF116847.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 795716 AK139455.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 795716 AK148073.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 795716 BC049776.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 795716 NM_019689.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 134910 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 239618 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 408158 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544399 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 771194 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 944255 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 633002 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 226032 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 282669 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193686 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 121137 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 452833 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 600650 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 137919 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 55812 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 91842 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154774 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153671 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2044653 AK028103.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2044653 BC060973.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2044653 NM_009999.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P472486 X99715.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P472486 AK028103.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P472486 BC060973.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P472486 NM_009999.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P472486 AF128849.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012395 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012381 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012401 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012400 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012403 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012382 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012390 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012385 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012383 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012380 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012396 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012387 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012379 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06144696 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01487594 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153671 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154774 - -
          Whole Mouse Genome G4122A A_52_P472486 X99715.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P472486 AK028103.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P472486 BC060973.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P472486 NM_009999.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P472486 AF128849.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P182362 M21856.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153671 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01487594 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154774 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06567882 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06567878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06221945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06263764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06567885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06257730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06567884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012401 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06454666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012445 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06257724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012396 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10551199 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551198 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551205 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551204 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551201 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551200 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551206 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551203 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551202 - -
          Affymetrix HT MG-430 PM Array Plate 1425645_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1422257_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1451787_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10551197 - -
          Mouse Genome 430 2.0 Array 1422257_s_at - -
          Mouse Genome 430 2.0 Array 1451787_PM_at - -
          Mouse Genome 430 2.0 Array 1425645_PM_s_at - -
          Mouse Genome 430 2.0 Array 1422257_PM_s_at - -
          Mouse Genome 430 2.0 Array 1425645_s_at - -
          Mouse Genome 430 2.0 Array 1451787_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13088_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13088_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700003347.mm.1 TC0700000430.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003347.mm.1 NM_009999.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003346.mm.1 TC0700000430.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003346.mm.1 NM_009999.4 Refseq
          Mouse Expression Set 430 1422257_s_at - -
          Mouse Expression Set 430 1422257_PM_s_at - -
          Murine Genome U74v2 Set 102701_PM_at - -
          Murine Genome U74v2 Set 102701_at - -
          Mouse Genome 430A 2.0 Array 1422257_s_at - -
          Mouse Genome 430A 2.0 Array 1422257_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1422257_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1451787_at - -
          Affymetrix HT Mouse Genome 430A Array 1425645_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1451787_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422257_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1425645_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6200139 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006200139 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1215955 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2594926 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215955 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2594926 - -
          mouse-6 v1.1 (Illumina) scl0013091.1_33-S - -
          Illumina Sentrix mouse-6.v1 bead array 6200139 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0013091.1_33-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6200139 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1215955 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2594926 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2594926 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0