Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01974474_gH
          See other RPLP0 GE Assays ›
          Gene Symbol:
          Rplp0
          Gene Name:
          ribosomal protein, large, P0
          Gene Aliases:
          36B4, Arbp, L10E
          Chromosome Location:
          Chr.5: 115559467 - 115563729 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01974474_gH
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rplp0
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007475.5 NP_031501.1 3 275 89
          GenBank mRNA AK002315.1 - 3 229 89
          AK010267.1 - 3 228 89
          AK012606.1 - 3 234 89
          AK029816.1 - 3 230 89
          AK081678.1 - 3 232 89
          AK150448.1 - 3 228 89
          AK159358.1 - 3 228 89
          AK160541.1 - 3 228 89
          AK161683.1 - 3 230 89
          AK166728.1 - 3 228 89
          AK167240.1 - 3 226 89
          AK191860.1 - 1 53 89
          AK192486.1 - 1 53 89
          BC003833.1 - 3 218 3600590 89
          BC057295.1 - 2 209 89
          BC087887.1 - 3 229 6594755 89
          BC089496.1 - 3 229 30254142 89
          BC099384.1 - 2 195 30843295 89
          X15267.1 - 3 227 89

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rplp0

          Panther Classification:

          Molecular Function -

          ribosomal protein translational protein

          Gene Ontology Categories:

          Function(s) Process(es)

          ribosome biogenesis
          cellular response to interleukin-4
          poly(A) RNA binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500010954.mm.1 TC0500001266.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500010954.mm.1 NM_007475.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010954.mm.1 NM_007475.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010956.mm.1 TC0500001266.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500010956.mm.1 NM_007475.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010956.mm.1 NM_007475.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010957.mm.1 TC0500001266.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500010957.mm.1 NM_007475.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010957.mm.1 NM_007475.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010952.mm.1 TC0500001266.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500010952.mm.1 NM_007475.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010952.mm.1 NM_007475.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010958.mm.1 TC0500001266.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500010958.mm.1 NM_007475.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010958.mm.1 NM_007475.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010943.mm.1 TC0500001266.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500010943.mm.1 NM_007475.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010943.mm.1 NM_007475.5 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 442133 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline