Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02002195_s1
          See other 6030471H07RIK GE Assays ›
          Gene Symbol:
          6030471H07Rik
          Gene Name:
          RIKEN cDNA 6030471H07 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02002195_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 6030471H07Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK020082.1 - 2 409 102

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 744447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03975537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065459 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065397 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03941903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03787548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065446 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065469 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065378 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065387 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03881152 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065458 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04046770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04046773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03772094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065381 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065461 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03725679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03744198 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03759256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03725668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03772088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03941904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03744195 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065392 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065376 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03744200 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065444 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23065355 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100460441 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline