Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02012635_s1
          See other OLFR94 GE Assays ›
          Gene Symbol:
          Olfr94
          Gene Name:
          olfactory receptor 94
          Gene Aliases:
          MOR256-32P
          Chromosome Location:
          Chr.17: 37196806 - 37197966 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02012635_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr94
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011518.1 NP_001011518.1 1 1119 72
          GenBank mRNA BC139173.1 - 1 1164 8862195 72
          BC139177.1 - 1 1164 8862199 72

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr94

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31167788 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31167791 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31167790 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31167787 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2014406 NM_001011518.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2014406 BC139173.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2014406 BC139177.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 614669 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 120487 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2014406 NM_001011518.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2014406 BC139173.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2014406 BC139177.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P313127 NM_001011518.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P313127 BC139173.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P313127 BC139177.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167788 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167787 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167822 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167807 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31167790 - -
          Whole Mouse Genome G4122A A_51_P313127 NM_001011518.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P313127 BC139173.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P313127 BC139177.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13195196 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13374612 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13262875 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31167791 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31167787 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31167788 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31167790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31167830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13354591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13210676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13421504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13126404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167779 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13421502 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13126402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167805 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13354596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31167837 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450863 - -
          Affymetrix Mouse Gene 1.1 ST Array 10450862 - -
          Affymetrix Mouse Clariom S Array PSR1700019543.mm.1 NM_001011518.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019543.mm.1 NM_001011518.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019543.mm.1 TC1700002058.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019544.mm.1 TC1700002058.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019542.mm.1 TC1700002058.mm.1 MTA TC
          Murine Genome U74v2 Set 102170_at - -
          Murine Genome U74v2 Set 102170_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 324675 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160479 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160479 - -
          mouse-6 v1.1 (Illumina) GI_58372121-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160479 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160479 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline