Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02393636_gH
          See other PRSS3 GE Assays ›
          Gene Symbol:
          Prss3
          Gene Name:
          protease, serine 3
          Gene Aliases:
          MTG, TRY4, Tb, Try3
          Chromosome Location:
          Chr.6: 41373759 - 41377613 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02393636_gH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prss3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011645.2 NP_035775.1 4 - 5 606 149
          GenBank mRNA AK131862.1 - 4 - 5 607 149
          BC098498.1 - 4 - 5 589 149

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prss3

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          digestion
          serine-type endopeptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P178679 BC098498.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P178679 NM_011645.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P178679 AK131862.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P394552 NM_001103153.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P394552 BC141304.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P394552 BC098498.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P394552 NM_011645.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P394552 AK131862.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24393631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24393650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24393663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24393635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24393664 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05875947 - -
          Whole Mouse Genome G4122A A_51_P394552 NM_001103153.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394552 BC141304.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394552 BC098498.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394552 NM_011645.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394552 AK131862.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P178679 BC098498.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P178679 NM_011645.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P178679 AK131862.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24393663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24393631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24393636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24393650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24393635 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10544346 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544339 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544345 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537653 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544343 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544340 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537654 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537652 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544344 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544341 - -
          Affymetrix HT MG-430 PM Array Plate 1450689_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10544338 - -
          Affymetrix Mouse Gene 1.1 ST Array 10537650 - -
          Affymetrix Mouse Gene 1.1 ST Array 10544342 - -
          Mouse Genome 430 2.0 Array 1450689_PM_at - -
          Mouse Genome 430 2.0 Array 1450689_at - -
          Mouse Genome 430 2.0 Array 1422682_PM_s_at - -
          Mouse Genome 430 2.0 Array 1422682_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22073_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22073_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600017005.mm.1 TC0600002155.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017005.mm.1 NM_011645.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017005.mm.1 NM_011645.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017000.mm.1 TC0600002155.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017000.mm.1 NM_011645.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017000.mm.1 NM_011645.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017003.mm.1 TC0600002155.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017003.mm.1 NM_011645.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017003.mm.1 NM_011645.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017002.mm.1 TC0600002155.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017002.mm.1 NM_011645.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017002.mm.1 NM_011645.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017001.mm.1 TC0600002155.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017001.mm.1 NM_011645.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017001.mm.1 NM_011645.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017004.mm.1 TC0600002155.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017004.mm.1 NM_011645.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017004.mm.1 NM_011645.2 Refseq
          Mouse Expression Set 430 1422682_s_at - -
          Mouse Expression Set 430 1422682_PM_s_at - -
          Mouse Expression Set 430 1450689_PM_at - -
          Mouse Expression Set 430 1450689_at - -
          Murine Genome U74v2 Set 93208_PM_at - -
          Murine Genome U74v2 Set 101339_at - -
          Murine Genome U74v2 Set 93208_at - -
          Murine Genome U74v2 Set 101339_PM_at - -
          Mouse Genome 430A 2.0 Array 1422682_s_at - -
          Mouse Genome 430A 2.0 Array 1450689_at - -
          Mouse Genome 430A 2.0 Array 1450689_PM_at - -
          Mouse Genome 430A 2.0 Array 1422682_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1450689_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450689_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 400820 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2896385 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2896385 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline