Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02524428_g1
          See other DEFA1 GE Assays ›
          Gene Symbol:
          Defa1
          Gene Name:
          defensin, alpha 1
          Gene Aliases:
          Defcr, Defcr1
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02524428_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Defa1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010031.2 NP_034161.2 2 364 58
          GenBank mRNA M33225.1 - 2 364 58
          X15617.1 - 2 364 58

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Defa1

          Panther Classification:

          Molecular Function -

          antimicrobial response protein

          Gene Ontology Categories:

          Function(s)

          defense response
          response to drug
          defense response to bacterium
          defense response to Gram-negative bacterium
          defense response to Gram-positive bacterium
          cellular response to lipopolysaccharide

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999108 NM_010031.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999108 M33225.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999110 U03061.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999110 NM_010031.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999110 M33225.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999110 X15617.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999110 U03061.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999110 NM_010031.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999110 M33225.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999110 X15617.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2068510 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999108 NM_010031.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999108 M33225.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P578763 NM_010031.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P578763 M33225.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P578763 X15617.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10570670 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570716 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570692 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570678 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570657 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570659 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570677 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570658 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570679 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570731 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570691 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570730 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570669 - -
          Affymetrix Mouse Gene 1.1 ST Array 10570711 - -
          Affymetrix Mouse Gene 1.1 ST Array 10570676 - -
          Affymetrix Mouse Gene 1.1 ST Array 10570656 - -
          Affymetrix Murine Genome 6500A Array Msa13720_at - -
          Murine Genome U74v2 Set 93879_f_at - -
          Murine Genome U74v2 Set 102814_PM_f_at - -
          Murine Genome U74v2 Set 93863_PM_f_at - -
          Murine Genome U74v2 Set 92812_PM_f_at - -
          Murine Genome U74v2 Set 95766_f_at - -
          Murine Genome U74v2 Set 93863_f_at - -
          Murine Genome U74v2 Set 95766_PM_f_at - -
          Murine Genome U74v2 Set 102814_f_at - -
          Murine Genome U74v2 Set 93879_PM_f_at - -
          Murine Genome U74v2 Set 92812_f_at - -
          Murine Genome U74v2 Set 167225_f_at - -
          Murine Genome U74v2 Set 167225_PM_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 545536 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4540619 - -
          MouseWG-6 v2 Expression BeadChip 6450019 - -
          Illumina mouse-6 v1.0 expression beadchip 4540619 - -
          Illumina mouse-6 v1.0 expression beadchip 6450019 - -
          Illumina mouse-6 v1.0 expression beadchip 1400601 - -
          Illumina mouse-6 v1.0 expression beadchip 101050609 - -
          Illumina mouse-6 v1.0 expression beadchip 1050152 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001050152 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004540619 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001400601 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006450019 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2731509 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2732554 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2669974 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1215673 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2669974 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252576 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2732554 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215673 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2731509 - -
          mouse-6 v1.1 (Illumina) scl0013230.1_27-S - -
          mouse-6 v1.1 (Illumina) scl0013216.1_7-S - -
          mouse-6 v1.1 (Illumina) scl0013229.1_27-S - -
          mouse-6 v1.1 (Illumina) GI_31981802-S - -
          mouse-6 v1.1 (Illumina) scl0023855.1_145-S - -
          Illumina Sentrix mouse-6.v1 bead array 1050152 - -
          Illumina Sentrix mouse-6.v1 bead array 1400601 - -
          Illumina Sentrix mouse-6.v1 bead array 101050609 - -
          Illumina Sentrix mouse-6.v1 bead array 6450019 - -
          Illumina Sentrix mouse-6.v1 bead array 4540619 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0023855.1_145-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0013230.1_27-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0013229.1_27-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0013216.1_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4540619 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1400601 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1050152 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101050609 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6450019 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2732554 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2669974 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2731509 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1215673 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2731509 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2732554 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2669974 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215673 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0023855.1_145-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0013216.1_7-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline