Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02525074_g1
          See other CSPRS GE Assays ›
          Gene Symbol:
          Csprs
          Gene Name:
          component of Sp100-rs
          Gene Aliases:
          D1Lub1, HSR
          Chromosome Location:
          Chr.1|NT_162750: 111572 - 163011 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02525074_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Csprs
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_033616.3 NP_291094.2 2 861 108
          GenBank mRNA AK037063.1 - 2 862 108

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Csprs

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2019601 BC016679.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2019601 BC038927.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2019601 NM_033616.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2019601 AK037063.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1973415 BC071209.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1973415 NM_033616.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1973415 AK037063.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063490 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2019601 BC016679.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2019601 BC038927.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2019601 NM_033616.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2019601 AK037063.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063490 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1973415 BC071209.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1973415 NM_033616.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1973415 AK037063.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P917015 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109245 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106632 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131841 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10582881 - -
          Affymetrix Mouse Gene 1.0 ST Array 10347916 - -
          Affymetrix Mouse Gene 1.0 ST Array 10347927 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582880 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356277 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356264 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356275 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356276 - -
          Affymetrix HT MG-430 PM Array Plate 1422141_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10356262 - -
          Affymetrix Mouse Gene 1.1 ST Array 10582879 - -
          Affymetrix Mouse Gene 1.1 ST Array 10347925 - -
          Affymetrix Mouse Gene 1.1 ST Array 10356274 - -
          Affymetrix Mouse Gene 1.1 ST Array 10347915 - -
          Mouse Genome 430 2.0 Array 1435792_PM_at - -
          Mouse Genome 430 2.0 Array 1422141_s_at - -
          Mouse Genome 430 2.0 Array 1422140_PM_at - -
          Mouse Genome 430 2.0 Array 1460416_s_at - -
          Mouse Genome 430 2.0 Array 1460416_PM_s_at - -
          Mouse Genome 430 2.0 Array 1435792_at - -
          Mouse Genome 430 2.0 Array 1422141_PM_s_at - -
          Mouse Genome 430 2.0 Array 1422140_at - -
          Affymetrix Mouse Clariom S Array PSR1_GL456221_random00000032.mm.1 NM_033616.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1_GL456221_random00000032.mm.1 TC1_GL456221_random00000019.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1_GL456221_random00000030.mm.1 NM_033616.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1_GL456221_random00000030.mm.1 TC1_GL456221_random00000019.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1_GL456221_random00000029.mm.1 NM_033616.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1_GL456221_random00000029.mm.1 TC1_GL456221_random00000019.mm.1 MTA TC
          Mouse Expression Set 430 1422140_PM_at - -
          Mouse Expression Set 430 1422141_s_at - -
          Mouse Expression Set 430 1422140_at - -
          Mouse Expression Set 430 1460416_s_at - -
          Mouse Expression Set 430 1422141_PM_s_at - -
          Mouse Expression Set 430 1460416_PM_s_at - -
          Murine Genome U74v2 Set 101846_PM_r_at - -
          Murine Genome U74v2 Set 101845_s_at - -
          Murine Genome U74v2 Set 101846_r_at - -
          Murine Genome U74v2 Set 93778_PM_at - -
          Murine Genome U74v2 Set 93778_at - -
          Murine Genome U74v2 Set 101845_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1460416_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1460416_s_at - -
          Mouse Genome 430A 2.0 Array 1422141_s_at - -
          Mouse Genome 430A 2.0 Array 1422140_at - -
          Mouse Genome 430A 2.0 Array 1422141_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1422140_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422141_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1422141_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 797753 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5860672 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005860672 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2661289 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2661289 - -
          mouse-6 v1.1 (Illumina) scl00114564.1_150-S - -
          Illumina Sentrix mouse-6.v1 bead array 5860672 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00114564.1_150-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5860672 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2661289 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2661289 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline