Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02525792_g1
          See other ZFP781 GE Assays ›
          Gene Symbol:
          Zfp781
          Gene Name:
          zinc finger protein 781
          Gene Aliases:
          A130081B01
          Chromosome Location:
          Chr.10: 81901911 - 81931231 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02525792_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp781
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_199062.1 NP_951017.1 4 3418 152
          GenBank mRNA BC062127.1 - 4 3418 6827033 152

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp781

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor zinc finger transcription factor C2H2 zinc finger transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999453 NM_199062.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999453 BC062127.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P580707 BC024063.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P580707 NM_199062.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P580707 BC062127.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P580707 BC024063.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P580707 NM_199062.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P580707 BC062127.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999453 NM_199062.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999453 BC062127.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P580707 BC024063.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P580707 NM_199062.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P580707 BC062127.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P580707 BC024063.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P580707 NM_199062.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P580707 BC062127.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09577459 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09577337 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09577300 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09577438 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07126397 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09577396 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09577330 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09577341 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09577415 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09577494 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09577407 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09577251 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09577371 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07126404 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09577511 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09577453 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112356 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09577310 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02052447 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09577257 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07126413 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09577315 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09577264 - -
          Whole Mouse Genome G4122A A_52_P580707 BC024063.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P580707 NM_199062.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P580707 BC062127.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P580707 BC024063.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P580707 NM_199062.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P580707 BC062127.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P207454 NM_199062.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P207454 AK038134.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P207454 BC062127.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136689 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09577371 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112356 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09577257 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07126413 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09577341 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09577438 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09577510 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09577459 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09577330 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09577494 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09577407 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09577264 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09577300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08262864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291273 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10371274 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371273 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365203 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371272 - -
          Affymetrix HT MG-430 PM Array Plate 1445274_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10365199 - -
          Affymetrix Mouse Gene 1.1 ST Array 10371271 - -
          Mouse Genome 430 2.0 Array 1446919_at - -
          Mouse Genome 430 2.0 Array 1446919_PM_at - -
          Mouse Genome 430 2.0 Array 1445274_PM_at - -
          Mouse Genome 430 2.0 Array 1445274_at - -
          Affymetrix Mouse Clariom S Array PSR1000019289.mm.1 NM_199062.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000019289.mm.1 TC1000002553.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000019286.mm.1 NM_199062.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000019286.mm.1 TC1000002553.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000019285.mm.1 NM_199062.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000019285.mm.1 TC1000002553.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000019284.mm.1 NM_199062.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000019284.mm.1 TC1000002553.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000019287.mm.1 NM_199062.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000019287.mm.1 TC1000002553.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000019290.mm.1 NM_199062.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000019290.mm.1 TC1000002553.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000019293.mm.1 NM_199062.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000019293.mm.1 TC1000002553.mm.1 MTA TC
          Mouse Expression Set 430 1445274_PM_at - -
          Mouse Expression Set 430 1445274_at - -
          Murine Genome U74v2 Set 135288_PM_at - -
          Murine Genome U74v2 Set 135288_at - -
          Affymetrix HT Mouse Genome 430B Array 1445274_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1445274_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101340546 - -
          Illumina mouse-6 v1.0 expression beadchip 101340546 - -
          Illumina mouse-6 v1.0 expression beadchip 106380538 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2612919 - -
          mouse-6 v1.1 (Illumina) scl00331188.1_102-S - -
          mouse-6 v1.1 (Illumina) scl0331188.1_6-S - -
          Illumina Sentrix mouse-6.v1 bead array 106380538 - -
          Illumina Sentrix mouse-6.v1 bead array 101340546 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106380538 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101340546 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2902928 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2660580 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2902928 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2660579 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline