Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02581281_m1
          See other RHOX10 GE Assays ›
          Gene Symbol:
          Rhox10
          Gene Name:
          reproductive homeobox 10
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 38066475 - 38071688 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02581281_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rhox10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024850.2 NP_001020021.1 1 - 2 474 78
          GenBank mRNA AK146090.1 - 1 - 2 456 78
          BC100366.1 - 1 - 2 418 30524902 78
          BC132474.1 - 1 - 2 405 40130751 78
          BC132500.1 - 1 - 2 405 40130777 78
          DQ058647.1 - 1 - 2 474 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00000359.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rhox10

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s)

          biological_process

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P138201 BC100366.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P138201 BC132474.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P138201 AK146090.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P138201 BC132500.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P138201 DQ058647.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928276 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 758369 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 475021 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 106122 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P138201 BC100366.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P138201 BC132474.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P138201 AK146090.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P138201 BC132500.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P138201 DQ058647.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326099 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326109 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326105 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326102 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32326095 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09267581 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15214116 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15214107 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15214110 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15214114 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15214105 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15214109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32326103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15000864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15105026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15147402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15125382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14728033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15105032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326098 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15125389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15144298 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15147401 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15125387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32326061 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10599315 - -
          Affymetrix Mouse Gene 1.0 ST Array 10599313 - -
          Affymetrix Mouse Gene 1.0 ST Array 10599312 - -
          Affymetrix Mouse Gene 1.0 ST Array 10599314 - -
          Affymetrix Mouse Gene 1.0 ST Array 10599311 - -
          Affymetrix Mouse Gene 1.0 ST Array 10599310 - -
          Affymetrix Mouse Gene 1.1 ST Array 10599309 - -
          Affymetrix Mouse Clariom S Array PSR0X00002463.mm.1 NM_001024850.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002463.mm.1 NM_001024850.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002463.mm.1 TC0X00000359.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00002466.mm.1 NM_001024850.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002466.mm.1 NM_001024850.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002466.mm.1 TC0X00000359.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00002467.mm.1 NM_001024850.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002467.mm.1 NM_001024850.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002467.mm.1 TC0X00000359.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00002465.mm.1 NM_001024850.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002465.mm.1 NM_001024850.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00002465.mm.1 TC0X00000359.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3162042 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3162042 - -
          mouse-6 v1.1 (Illumina) GI_71480139-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3162042 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3162042 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline