Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02581594_m1
          See other PRR27 GE Assays ›
          Gene Symbol:
          Prr27
          Gene Name:
          proline rich 27
          Gene Aliases:
          4930432K09Rik
          Chromosome Location:
          Chr.5: 87825697 - 87846386 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02581594_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prr27
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163551.1 NP_001157023.1 4 - 5 305 107
          NM_028648.3 NP_082924.2 3 - 4 281 83
          GenBank mRNA AK015291.1 - 3 - 4 282 83
          BC103787.1 - 4 - 5 316 30919261 107
          BY714849.1 - 3 - 4 282 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000798.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prr27

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071008 AK015291.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071008 NM_028648.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071008 BC049645.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071008 BC103787.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071008 NM_001163551.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 741752 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 146676 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 144456 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 317915 AK036572.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 317915 AK051050.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 317915 AK156604.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 317915 BC065139.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 317915 NM_008396.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 317915 Z29987.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 525163 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 466191 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 58592 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 6771 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 164593 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 71350 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 611504 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 18612 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 173840 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 137011 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155933 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P144333 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071008 AK015291.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071008 NM_028648.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071008 BC049645.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071008 BC103787.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071008 NM_001163551.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23869742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23869748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23869750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23869845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23869838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23869745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23869739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23869735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23869744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23869839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23869752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23869740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23869837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23869840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23869734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23869843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23869738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23869835 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05645656 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155933 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01154444 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P144333 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05645654 - -
          Whole Mouse Genome G4122A A_51_P464691 AK015291.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P464691 NM_028648.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P464691 BC049645.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P464691 BC103787.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P464691 NM_001163551.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155933 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P144333 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04989861 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04989867 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04989863 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04749805 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04989864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23869911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869899 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869883 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04384935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04812612 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869820 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04875571 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04200170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869901 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869738 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04845358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04755289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04200166 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04356360 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869753 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04890845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04783008 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869907 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869876 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869868 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869851 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869823 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869873 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04875572 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869894 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04200171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23869735 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10522872 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522871 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522869 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522870 - -
          Affymetrix HT MG-430 PM Array Plate 1431447_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10522868 - -
          Mouse Genome 430 2.0 Array 1431447_at - -
          Mouse Genome 430 2.0 Array 1431447_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73779_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73779_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500007009.mm.1 TC0500000798.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007009.mm.1 NM_001163551.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007009.mm.1 NM_001163551.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007009.mm.1 NM_028648.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007009.mm.1 NM_028648.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007008.mm.1 TC0500000798.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007008.mm.1 NM_001163551.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007008.mm.1 NM_001163551.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007008.mm.1 NM_028648.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007008.mm.1 NM_028648.3 Refseq
          Mouse Expression Set 430 1431447_at - -
          Mouse Expression Set 430 1431447_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431447_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431447_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2850040 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2760525 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2760525 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2760525 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline