Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02600438_m1
          See other KMT2D GE Assays ›
          Gene Symbol:
          Kmt2d
          Gene Name:
          lysine (K)-specific methyltransferase 2D
          Gene Aliases:
          ALR, BC032281, BC058659, C430014K11Rik, KMT2B, Mll2, Mll4
          Chromosome Location:
          Chr.15: 98831669 - 98871205 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02600438_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kmt2d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033276.3 NP_001028448.3 15 - 16 4328 62
          GenBank mRNA AK140390.1 - 6 - 7 2906 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500002192.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          oocyte growth
          in utero embryonic development
          chromatin silencing
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          positive regulation of cell proliferation
          chromatin modification
          methylation
          positive regulation of intracellular estrogen receptor signaling pathway
          response to estrogen
          positive regulation of transcription from RNA polymerase II promoter
          oogenesis
          histone H3-K4 methylation
          methyltransferase activity
          zinc ion binding
          transferase activity
          histone-lysine N-methyltransferase activity
          transcription regulatory region DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477060 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477062 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477055 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477058 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477059 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477063 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477057 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477061 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136533 BC058659.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136533 NM_001033276.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136526 BC058659.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136526 NM_001033276.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967733 NM_001033276.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967733 AK140263.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 614194 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 537846 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 9877 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175295 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115519 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136526 BC058659.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136526 NM_001033276.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136533 BC058659.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136533 NM_001033276.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967733 NM_001033276.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967733 AK140263.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175295 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115519 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03112299 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03112308 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102065 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03112299 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115519 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175295 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477058 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477055 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477062 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477061 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477057 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477060 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477059 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30477063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30476814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12375811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12109714 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30476992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12375810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30476843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30476834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30476828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30476999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30476831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30476846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30476822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30476996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30476841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11911856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30476824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30476839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30476829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30476990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30476817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30476994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30476844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12109733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30476836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30477019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12375801 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10432312 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432301 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432308 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432348 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432306 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432311 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432310 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432300 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432307 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432309 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432313 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432297 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432303 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432305 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432299 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432302 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432295 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432304 - -
          Affymetrix HT MG-430 PM Array Plate 1427555_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1435550_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10432294 - -
          Affymetrix Mouse Gene 1.1 ST Array 10432298 - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 381022_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 381022_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500017895.mm.1 NM_001033276.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017895.mm.1 NM_001033276.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500017895.mm.1 TC1500002192.mm.1 MTA TC
          Affymetrix HT Mouse Genome 430A Array 1427555_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427555_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 611064 - -
          ABI Mouse Genome Survey Microarray 708372 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3162228 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3162228 - -
          mouse-6 v1.1 (Illumina) GI_85701621-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline