Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02746179_m1
          See other BC055324 GE Assays ›
          Gene Symbol:
          BC055324
          Gene Name:
          cDNA sequence BC055324
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 163945840 - 163994900 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02746179_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BC055324
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_201364.1 NP_958752.1 9 - 10 1013 76
          GenBank mRNA AK031594.1 - 9 - 10 1009 76
          AK141661.1 - 9 - 10 1017 76
          AK163374.1 - 917 76
          BC052855.1 - 9 - 10 1007 30058659 76
          BC055324.1 - 9 - 10 1013 5715852 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003416.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene BC055324

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100028052.mm.1 NM_201364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028052.mm.1 NM_201364.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028052.mm.1 TC0100003416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028046.mm.1 NM_201364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028046.mm.1 NM_201364.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028046.mm.1 TC0100003416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028057.mm.1 NM_201364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028057.mm.1 NM_201364.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028057.mm.1 TC0100003416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028039.mm.1 NM_201364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028039.mm.1 NM_201364.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028039.mm.1 TC0100003416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028045.mm.1 NM_201364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028045.mm.1 NM_201364.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028045.mm.1 TC0100003416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028051.mm.1 NM_201364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028051.mm.1 NM_201364.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028051.mm.1 TC0100003416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028063.mm.1 NM_201364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028063.mm.1 NM_201364.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028063.mm.1 TC0100003416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028049.mm.1 NM_201364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028049.mm.1 NM_201364.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028049.mm.1 TC0100003416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028056.mm.1 NM_201364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028056.mm.1 NM_201364.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028056.mm.1 TC0100003416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028042.mm.1 NM_201364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028042.mm.1 NM_201364.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028042.mm.1 TC0100003416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028044.mm.1 NM_201364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028044.mm.1 NM_201364.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028044.mm.1 TC0100003416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028054.mm.1 NM_201364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028054.mm.1 NM_201364.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028054.mm.1 TC0100003416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028059.mm.1 NM_201364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028059.mm.1 NM_201364.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028059.mm.1 TC0100003416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028061.mm.1 NM_201364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028061.mm.1 NM_201364.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028061.mm.1 TC0100003416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028041.mm.1 NM_201364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028041.mm.1 NM_201364.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028041.mm.1 TC0100003416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028048.mm.1 NM_201364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028048.mm.1 NM_201364.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028048.mm.1 TC0100003416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028050.mm.1 NM_201364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028050.mm.1 NM_201364.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028050.mm.1 TC0100003416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028053.mm.1 NM_201364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028053.mm.1 NM_201364.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028053.mm.1 TC0100003416.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 311449 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline