Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03012652_s1
          See other OLFR532 GE Assays ›
          Gene Symbol:
          Olfr532
          Gene Name:
          olfactory receptor 532
          Gene Aliases:
          MOR251-1
          Chromosome Location:
          Chr.7: 140418842 - 140419771 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03012652_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr532
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147026.1 NP_667237.1 1 438 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004501.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr532

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P132621 NM_147026.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P144036 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P132621 NM_147026.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583712 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583724 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P144036 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P144036 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06420928 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06420938 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06420912 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06420918 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06420935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06420910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06420926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06659053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06466860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06466863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583716 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10568963 - -
          Affymetrix Mouse Gene 1.1 ST Array 10568962 - -
          Affymetrix Mouse Clariom S Array PSR0700037813.mm.1 NM_147026.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037813.mm.1 NM_147026.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037813.mm.1 TC0700004501.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218630 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2895651 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2895651 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline