Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03013503_g1
          See other OLFR663 GE Assays ›
          Gene Symbol:
          Olfr663
          Gene Name:
          olfactory receptor 663
          Gene Aliases:
          MOR40-12
          Chromosome Location:
          Chr.7: 104703569 - 104704603 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03013503_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr663
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011757.1 NP_001011757.1 1 979 73

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr663

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163468 NM_001011757.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 17228 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163468 NM_001011757.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06382375 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05948629 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05948620 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05948630 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05948623 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05948627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06063560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05948629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06365889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06189846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06536945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06602721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389261 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555891 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555890 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555889 - -
          Affymetrix Mouse Clariom S Array PSR0700012911.mm.1 TC0700001533.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012911.mm.1 NM_001011757.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012911.mm.1 NM_001011757.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012912.mm.1 TC0700001533.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012912.mm.1 NM_001011757.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012912.mm.1 NM_001011757.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 712337 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160944 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160944 - -
          mouse-6 v1.1 (Illumina) GI_58801301-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160944 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160944 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline