Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03013513_s1
          See other OLFR299 GE Assays ›
          Gene Symbol:
          Olfr299
          Gene Name:
          olfactory receptor 299
          Gene Aliases:
          MOR221-2
          Chromosome Location:
          Chr.7: 86465413 - 86466405 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03013513_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr299
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011767.1 NP_001011767.1 1 549 125

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr299

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P458707 NM_001011767.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P458707 NM_001011767.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 877604 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P458707 NM_001011767.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P458707 NM_001011767.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P458707 NM_001011767.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P458707 NM_001011767.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294788 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294807 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25294809 - -
          Whole Mouse Genome G4122A A_51_P458707 NM_001011767.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P458707 NM_001011767.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06100314 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06595506 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06595518 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06151839 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06100313 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06595515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25294818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06566809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06595525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06548399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25294784 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10554726 - -
          Affymetrix Mouse Gene 1.1 ST Array 10554725 - -
          Affymetrix Mouse Clariom S Array PSR0700010770.mm.1 TC0700001241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700010770.mm.1 NM_001011767.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010770.mm.1 NM_001011767.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 687236 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2998284 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2998284 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0