Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03015334_m1
          See other GM525 GE Assays ›
          Gene Symbol:
          Gm525
          Gene Name:
          predicted gene 525
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 89073841 - 89093064 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03015334_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm525
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033266.2 NP_001028438.1 3 - 4 399 88
          GenBank mRNA AA237157.1 - 3 - 4 295 88
          AK040024.1 - 3 - 4 317 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001389.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017977 NM_001033266.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017977 AA237157.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017977 AK040024.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 292232 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 783854 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 740035 AK016080.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 856243 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 482106 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 293225 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 604485 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 384791 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 826006 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 4488 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 145104 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017977 NM_001033266.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017977 AA237157.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017977 AK040024.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02389967 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02389952 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07435355 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02389967 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02389952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09070457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09613203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09423107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09223174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09277518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09231267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09406391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09493164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08989663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09070452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09223179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09070456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09303646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09016341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09668024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09613210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09261989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09406392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09601110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09668023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09531624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09493163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09550159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09339589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003652 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10380281 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380282 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380284 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380283 - -
          Affymetrix Mouse Gene 1.1 ST Array 10380280 - -
          Affymetrix Mouse Clariom S Array PSR1100013034.mm.1 NM_001033266.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013034.mm.1 NM_001033266.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013034.mm.1 TC1100001389.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100013033.mm.1 NM_001033266.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013033.mm.1 NM_001033266.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013033.mm.1 TC1100001389.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253832 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3137124 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3059859 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3137124 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3059859 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline