Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03015615_s1
          See other KRTAP5-5 GE Assays ›
          Gene Symbol:
          Krtap5-5
          Gene Name:
          keratin associated protein 5-5
          Gene Aliases:
          A030001H12Rik, AI586182, BB149782
          Chromosome Location:
          Chr.7: 142228795 - 142229971 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03015615_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Krtap5-5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037822.1 NP_001032911.1 1 1122 71
          GenBank mRNA AK020690.1 - 3 943 71
          BC111113.1 - 1 1122 40051363 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004559.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Krtap5-5

          Gene Ontology Categories:

          Function(s) Process(es)

          hematopoietic progenitor cell differentiation
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2060011 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P101703 NM_001037822.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P101703 AK020690.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P101703 BC111113.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1985294 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098150 NM_001037822.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098150 BC111113.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098150 NM_001037822.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098150 BC111113.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1985294 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P101703 NM_001037822.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P101703 AK020690.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P101703 BC111113.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2060011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593477 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593457 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25593456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25593427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05910838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05946636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06245541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06470526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06016783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06183824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06307425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06470528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06329823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06307428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06559077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06358891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25593449 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10559176 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569307 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569301 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569302 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569289 - -
          Affymetrix HT MG-430 PM Array Plate 1430728_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10569306 - -
          Affymetrix Mouse Gene 1.1 ST Array 10569300 - -
          Affymetrix Mouse Gene 1.1 ST Array 10559175 - -
          Mouse Genome 430 2.0 Array 1430728_PM_at - -
          Mouse Genome 430 2.0 Array 1430728_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 114666_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 114666_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700038438.mm.1 NM_001037822.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038438.mm.1 NM_001037822.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038438.mm.1 TC0700004559.mm.1 MTA TC
          Affymetrix HT Mouse Genome 430B Array 1430728_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430728_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 443209 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_3007540 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3007540 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline