Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03038613_g1
          See other VMN2R84 GE Assays ›
          Gene Symbol:
          Vmn2r84
          Gene Name:
          vomeronasal 2, receptor 84
          Gene Aliases:
          EG625068
          Chromosome Location:
          Chr.10: 130385800 - 130394241 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03038613_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn2r84
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081448.1 NP_001074917.1 3 - 4 1293 91

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 843453 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 177386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27549597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27549605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27549584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27549613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27549586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27549624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27549610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27549588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27549599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27549602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27549603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27549612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27549608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27549676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27549645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27549600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27549649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27549596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27549615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27549609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27549590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27549591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08723130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27549695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27549649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27549639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08422119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27549676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27549645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08405783 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10405871 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373688 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373686 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410206 - -
          Affymetrix Mouse Gene 1.0 ST Array 10405884 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373689 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373687 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410253 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373685 - -
          Affymetrix HT MG-430 PM Array Plate 1442282_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10405870 - -
          Affymetrix Mouse Gene 1.1 ST Array 10410204 - -
          Affymetrix Mouse Gene 1.1 ST Array 10410252 - -
          Affymetrix Mouse Gene 1.1 ST Array 10405879 - -
          Affymetrix Mouse Gene 1.1 ST Array 10373684 - -
          Mouse Genome 430 2.0 Array 1442282_at - -
          Mouse Genome 430 2.0 Array 1442282_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000023999.mm.1 NM_001081448.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023999.mm.1 NM_001081448.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023999.mm.1 TC1000003229.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000024000.mm.1 NM_001081448.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000024000.mm.1 NM_001081448.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000024000.mm.1 TC1000003229.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000024005.mm.1 NM_001081448.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000024005.mm.1 NM_001081448.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000024005.mm.1 TC1000003229.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000024006.mm.1 NM_001081448.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000024006.mm.1 NM_001081448.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000024006.mm.1 TC1000003229.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000024002.mm.1 NM_001081448.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000024002.mm.1 NM_001081448.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000024002.mm.1 TC1000003229.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000024004.mm.1 NM_001081448.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000024004.mm.1 NM_001081448.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000024004.mm.1 TC1000003229.mm.1 MTA TC
          Mouse Expression Set 430 1442282_at - -
          Mouse Expression Set 430 1442282_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1442282_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1442282_at - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2918728 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2918728 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline