Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03039691_s1
          See other OLFR430 GE Assays ›
          Gene Symbol:
          Olfr430
          Gene Name:
          olfactory receptor 430
          Gene Aliases:
          MOR105-5P
          Chromosome Location:
          Chr.1: 174069300 - 174070253 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03039691_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr430
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146718.2 NP_666929.2 1 906 100
          GenBank mRNA BC106773.1 - 1 851 100
          BC106774.1 - 1 848 100
          BC126860.1 - 1 845 100
          BC126861.1 - 1 845 100

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr430

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957695 BC126861.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957695 NM_146718.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957695 BC106773.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957695 BC106774.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120468 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957695 BC126861.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957695 NM_146718.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957695 BC106773.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957695 BC106774.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P480427 BC126860.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20840682 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120468 - -
          Whole Mouse Genome G4122A A_51_P480427 BC126860.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140001 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120468 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00938119 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00938098 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00938110 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00938101 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00938108 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00938097 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00938114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20840664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00938109 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00938106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00795529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00791958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00938119 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00938111 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00756947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00938110 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840702 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00383603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00938112 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00938098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20840681 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10351889 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351890 - -
          Affymetrix Mouse Gene 1.1 ST Array 10351888 - -
          Affymetrix Mouse Clariom S Array PSR0100013614.mm.1 NM_146718.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013614.mm.1 NM_146718.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013614.mm.1 TC0100001638.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 624878 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3780563 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003780563 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1245085 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245085 - -
          mouse-6 v1.1 (Illumina) scl14322.1.1_227-S - -
          Illumina Sentrix mouse-6.v1 bead array 3780563 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl14322.1.1_227-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3780563 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2614255 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2614255 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline