Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03047641_s1
          See other OLFR983 GE Assays ›
          Gene Symbol:
          Olfr983
          Gene Name:
          olfactory receptor 983
          Gene Aliases:
          MOR162-1
          Chromosome Location:
          Chr.9: 40092017 - 40093558 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03047641_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr983
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146827.2 NP_667038.1 2 559 82

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr983

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P420577 NM_146827.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P420577 NM_146827.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175580 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P420577 NM_146827.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P420577 NM_146827.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P420577 NM_146827.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P420577 NM_146827.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441295 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441255 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441279 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26441280 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175580 - -
          Whole Mouse Genome G4122A A_51_P420577 NM_146827.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P420577 NM_146827.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111052 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175580 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08109231 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08109268 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08109269 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08109265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26441278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07909345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08109248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07811133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07523368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07712486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07811138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07811141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08109239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07468410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26441240 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10592466 - -
          Affymetrix Mouse Gene 1.1 ST Array 10592465 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 890713 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2810364 - -
          Illumina mouse-6 v1.0 expression beadchip 2810364 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002810364 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1256987 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256987 - -
          mouse-6 v1.1 (Illumina) scl36005.1.254_69-S - -
          Illumina Sentrix mouse-6.v1 bead array 2810364 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36005.1.254_69-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2810364 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1256987 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1256987 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline