Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03049152_s1
          See other OLFR1052 GE Assays ›
          Gene Symbol:
          Olfr1052
          Gene Name:
          olfactory receptor 1052
          Gene Aliases:
          MOR172-1
          Chromosome Location:
          Chr.2: 86297818 - 86298756 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03049152_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1052
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147010.2 NP_667221.2 1 623 91
          GenBank mRNA BC127979.1 - 1 627 40135213 91

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1052

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          response to stimulus
          detection of chemical stimulus involved in sensory perception of smell
          signal transducer activity
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982643 NM_147010.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982643 BC127979.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2007185 NM_147010.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2007185 BC127979.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171601 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2007185 NM_147010.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2007185 BC127979.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982643 NM_147010.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982643 BC127979.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P356751 NM_147010.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21401647 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171601 - -
          Whole Mouse Genome G4122A A_51_P356751 NM_147010.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102771 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171601 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01772174 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01772176 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01772167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401629 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401657 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01441381 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401628 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01204593 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02036693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01441383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401683 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01907728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401642 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473504 - -
          Affymetrix Mouse Gene 1.1 ST Array 10473503 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6020398 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006020398 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2769198 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2769198 - -
          mouse-6 v1.1 (Illumina) scl4926.1.1_78-S - -
          Illumina Sentrix mouse-6.v1 bead array 6020398 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4926.1.1_78-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6020398 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3006729 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3006729 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2769198 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0